Skip to Content
Merck
All Photos(1)

Key Documents

H1429400

Hydroxyethyl salicylate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ethylene glycol monosalicylate, Ethylene glycol salicylate, Salicylic acid 2-hydroxyethyl ester, Salicylic acid ethylene glycol ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COOCH2CH2OH
CAS Number:
Molecular Weight:
182.17
Beilstein:
2255685
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hydroxyethyl salicylate

manufacturer/tradename

EDQM

refractive index

n20/D 1.548 (lit.)

bp

166 °C/13 mmHg (lit.)

density

1.244 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OCCOC(=O)c1ccccc1O

InChI

1S/C9H10O4/c10-5-6-13-9(12)7-3-1-2-4-8(7)11/h1-4,10-11H,5-6H2

InChI key

LVYLCBNXHHHPSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydroxyethyl salicylate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contact dermatitis from anti-inflammatory gel containing hydroxyethyl salicylate.
J Horak et al.
Contact dermatitis, 47(2), 120-121 (2002-11-29)
Miroslav Kucera et al.
Arzneimittel-Forschung, 53(12), 850-856 (2004-01-22)
A combination of the active agents arnica and hydroxyethyl salicylate (HES) in ethanolic solution (Sportino Acute Spray) is cutaneously applied for the treatment of sports injuries and diseases of the locomotor apparatus. The aim was to examine the efficacy and
Contact dermatitis from hydroxyethyl salicylate.
C Reichert et al.
Contact dermatitis, 33(4), 275-276 (1995-10-01)
Allergic contact dermatitis from a veterinary antiinflammatory gel containing 2-hydroxyethyl salicylate.
Susana Córdoba et al.
Dermatitis : contact, atopic, occupational, drug, 22(3), 171-172 (2011-05-17)
S E Cross et al.
British journal of clinical pharmacology, 46(1), 29-35 (1998-08-05)
The penetration of active ingredients from topically applied anti-inflammatory pharmaceutical products into tissues below the skin is the basis of their therapeutic efficacy. There is still controversy as to whether these agents are capable of direct penetration by diffusion through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service