Skip to Content
Merck
All Photos(3)

Documents

E4009

Sigma-Aldrich

Eosin Y

Dye content ~99%, Powder

Synonym(s):

2′,4′,5′,7′-Tetrabromofluorescein, Acid Red 87, Bromo acid J. TS, XL, or XX, Bromofluorescein, Bronze Bromo ES, Eosin yellowish, Solvent red 43

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H8Br4O5
CAS Number:
Molecular Weight:
647.89
Colour Index Number:
45380:2
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Eosin Y, Dye content ~99 %

form

powder

Quality Level

composition

Dye content, ~99%

color

light red to red/pink

solubility

ethanol: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1c(Br)cc2c(Oc3c(Br)c(O)c(Br)cc3C24OC(=O)c5ccccc45)c1Br

InChI

1S/C20H8Br4O5/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20/h1-6,25-26H

InChI key

DBZJJPROPLPMSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Eosin Y has been used as a stain:
  • to determine the location of the brain sections
  • for staining of fixed liver tissue
  • for histological examination of brain sections from the dorsal hippocampus

Biochem/physiol Actions

Eosin Y is a xanthene dye and is used for the differential staining of connective tissue and cytoplasm. In histopathology, it is applied as a counterstain after hematoxylin and before methylene blue. It is also used as a background stain, thereby giving contrast to the nuclear stains.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonello Mai et al.
Journal of medicinal chemistry, 51(7), 2279-2290 (2008-03-20)
A number of new compounds bearing two ortho-bromo- and ortho, ortho-dibromophenol moieties linked through a saturated/unsaturated, linear/(poly)cyclic spacer (compounds 1- 9) were prepared as simplified analogues of AMI-5 (eosin), a recently reported inhibitor of both protein arginine and histone lysine
An immunohistochemical analysis of SERT in the blood-brain barrier of the male rat brain.
Young LW, et al.
Histochemistry and Cell Biology, 144, 321-321 (2015)
Florian Ménard et al.
Bioorganic & medicinal chemistry, 17(22), 7647-7657 (2009-10-13)
This paper reports the synthesis and characterization of a new class of tetrabenzoporphyrins bearing glucosyl or polyamine units on meso positions to improve the targeting of cancer cells. Photocytotoxic activity of these photosensitizers was tested on cell lines HaCaT and
Inactivation of NF-kappaB p50 leads to insulin sensitization in liver through post-translational inhibition of p70S6K.
Gao Z, et al.
The Journal of Biological Chemistry, 284, 18368-18368 (2009)
The effects of simvastatin on hippocampal caspase-3 and Bcl-2 expression following kainate-induced seizures in rats.
Sun J, et al.
International Journal of Molecular Medicine, 30, 739-739 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service