Skip to Content
Merck
All Photos(1)

Key Documents

CG50

Sigma-Aldrich

Amberlite CG50

macroreticular

Synonym(s):

Amberlite Weakly Acidic Cation Exchanger, Hydrogen form

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

form

macroreticular

Quality Level

parameter

≤120 °C

technique(s)

activity assay: suitable

matrix

4% cross-linked methacrylate

matrix activation

carboxylic acid

particle size

100-200 mesh (wet)

pH

5-14(effective)

capacity

10.0 meq/g
3.5 meq/mL

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite is an ion exchange resin, which is macroporous weak acid cation exchange resin. The matrix is a polyacrylic with a particle diameter of 500 − 750 μm.

Application

Amberlite Weakly Acidic Cation Exchanger, Hydrogen form has been used in purification of thiamine from crude food extracts and bovine lactoperoxidase enzyme.
The Amberlite weakly acidic cation exchanger is in the hydrogen form and is used in protein chromatography, ion exchange chromatography and cation exchange media.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Ho Kim et al.
Biomolecules & therapeutics, 21(3), 234-240 (2013-11-23)
Monoamine oxidase inhibitors (MAOI) have been widely used as antidepressants. Recently, there has been renewed interest in MAO inhibitors. The activity-guided fractionation of extracts from Angelica keiskei Koidzumi (A. keiskei K.) led to the isolation of two prenylated chalcones, xanthoangelol
Kinetic parameters of thiamine degradation in NASA spaceflight foods determined by the endpoints method for long-term storage
Goulette TR, et al.
Food Chemistry, 302, 125365-125365 (2020)
Mohammad Said Hosseini et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 19(11), 1505-1508 (2003-12-03)
A method for the trace amount determination of Tl(I), via its preconcentration, is proposed. The method is based on the reaction of iodide, Tl(I) and Rhodamine B in a weakly acidic medium. In this process an ion-associated complex is formed
Martin Raithel et al.
BMC gastroenterology, 15, 41-41 (2015-04-19)
Patients with gastrointestinal food allergy are characterised by increased production of mast cell derived mediators upon allergen contact and present often with unspecific symptoms. The aim of this study was to evaluate urinary histamine and methylhistamine excretion in patients with
Timothy R Goulette et al.
Food chemistry, 302, 125365-125365 (2019-08-24)
Retention of labile vitamins such as thiamine (vitamin B1) in NASA spaceflight foods intended for extended-duration missions is critical for the health of the crew. In this study, the degradation kinetics of thiamine in three NASA spaceflight foods (brown rice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service