Skip to Content
Merck
All Photos(1)

Key Documents

A0325200

Alcuronium chloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N,N′-Diallylnortoxiferinium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H50Cl2N4O2
CAS Number:
Molecular Weight:
737.80
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

alcuronium

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[Cl-].[Cl-].[N+]21([C@@H]3C4([C@H]5N(c%11c4cccc%11)\C=C6/[C@@H]7N(c8c(cccc8)C97[C@H]%10[N+](CC9)(C\C(=C\CO)\C/6C%10)CC=C)\C=C/5\C(C3)\C(=C/CO)\C2)CC1)CC=C

InChI

1S/C44H50N4O2.2ClH/c1-3-17-47-19-15-43-35-9-5-7-11-37(35)45-26-34-32-24-40-44(16-20-48(40,18-4-2)28-30(32)14-22-50)36-10-6-8-12-38(36)46(42(34)44)25-33(41(43)45)31(23-39(43)47)29(27-47)13-21-49;;/h3-14,25-26,31-32,39-42,49-50H,1-2,15-24,27-28H2;2*1H/q+2;;/p-2/b29-13-,30-14-,33-25-,34-26-;;/t31?,32?,39-,40-,41-,42-,43?,44?,47?,48?;;/m0../s1

InChI key

CPYGBGOXCJJJGC-ZHNYPADJSA-L

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alcuronium chloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Christopoulos et al.
Biochemical Society transactions, 32(Pt 5), 873-877 (2004-10-21)
Allosteric modulators of G-protein-coupled receptors interact with binding sites that are topographically distinct from the orthosteric site recognized by the receptor's endogenous agonist. Allosteric ligands offer a number of advantages over orthosteric drugs, including the potential for greater receptor subtype
A Schröter et al.
Naunyn-Schmiedeberg's archives of pharmacology, 362(6), 512-519 (2001-01-04)
Muscarinic M2 acetylcholine receptors contain an allosteric site that is probably located at the entrance of the ligand binding pocket above the orthosteric binding site. With the orthosteric area not occupied, allosteric agents might gain access to this site. The
J Jakubík et al.
Molecular pharmacology, 54(5), 899-906 (1998-11-06)
We investigated whether alcuronium, an allosteric modulator of muscarinic acetylcholine receptors, can induce receptor-mediated activation of Go proteins in liposomal membranes incorporating purified M2 receptors and Go proteins and whether its action is affected by the receptor/Go protein (R/Go) ratio.
U Holzgrabe et al.
Journal of chemical information and computer sciences, 36(5), 1018-1024 (1996-09-01)
Structurally dissimilar compounds such as alcuronium and the newly synthesized substances derived from the bisbenzyl ether TMB4 and from hexamethonium stabilize antagonist binding to M2-cholinoceptors which is indicative of an allosteric action. In order to propose a hypothesis for the
J Jakubík et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(16), 8705-8709 (1996-08-06)
Ligands that bind to the allosteric-binding sites on muscarinic acetylcholine receptors alter the conformation of the classical-binding sites of these receptors and either diminish or increase their affinity for muscarinic agonists and classical antagonists. It is not known whether the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service