Skip to Content
Merck
All Photos(1)

Key Documents

93313

Supelco

Trizma® hydrochloride buffer solution

BioUltra, for molecular biology, pH 7.4

Synonym(s):

Tris(hydroxymethyl)aminomethane hydrochloride buffer solution

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
3675235
MDL number:
UNSPSC Code:
12000000
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
ND.02

grade

for molecular biology

Quality Level

product line

BioUltra

form

liquid

concentration

1.0 M tris-HCl

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

7.4±0.1 (25 °C)

density

1.05 g/mL at 20 °C

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.04

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Reduced glutathione inhibits rabbit and rat skeletal muscle lactate dehydrogenase and prevents dinitrophenol induced extracellular acidification by an epithelial cell line.: This study investigates how Trizma® hydrochloride buffer solution maintains pH stability, critical for accurately assessing the biochemical interactions in muscle and epithelial cells (Wimberger et al., 1997).

Other Notes

Buffer component (SET buffer) in the preparation of RNAcitation
Buffer component (SET buffer) in the preparation of RNA

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation of RNA.
S L Berger et al.
Methods in enzymology, 180, 3-13 (1989-01-01)
Anna Iwaniak et al.
International journal of molecular sciences, 22(6) (2021-04-04)
In silico and in vitro methods were used to analyze ACE- and DPP-IV-inhibiting potential of Gouda cheese with a modified content of β-casein. Firstly, the BIOPEP-UWM database was used to predict the presence of ACE and DPP-IV inhibitors in casein
Satish N Dighe et al.
ACS omega, 5(48), 30971-30979 (2020-12-17)
Cholinesterases (ChE) are well-known drug targets for the treatment of Alzheimer's disease (AD). In continuation of work to develop novel cholinesterase inhibitors, we utilized a structure-based scaffold repurposing approach and discovered six novel ChE inhibitors from our recently developed DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service