Skip to Content
Merck
All Photos(1)

Documents

91554

Supelco

4-Hydroxybenzaldehyde

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CHO
CAS Number:
Molecular Weight:
122.12
Beilstein:
471352
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

112-116 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)c1ccc(O)cc1

InChI

1S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9H

InChI key

RGHHSNMVTDWUBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxybenzaldehyde is a polar aromatic compound, reported to be one of the principal flavor components, contributing to the vanilla flavor, in the food and beverage industry. It is reported to be found in pea protein and identified in plant materials, such as vanilla beans as a product of the enzymatic reaction pathway.

Application

4-Hydroxybenzaldehyde may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Natural vanilla extracts and synthetic flavoring using micellar electrokinetic capillary chromatography (MECC).
  • Natural vanilla extracts and vanilla-flavored foods using thin layer chromatography (TLC) on silica gel and automated multiple development.
  • Pea protein UHT beverages using gas chromatography/mass spectrometry/olfactometry (GC/MS/O), further quantified by dynamic headspace and tandem gas chromatography/mass spectrometry (DHS-GC/MS-QQQ).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

345.2 °F

Flash Point(C)

174 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of aroma compounds in pea protein UHT beverages
Trikusuma M, et al.
Food Chemistry, 312 (2020)
Preparation and antitumor effects of 4-amino-1,2,4-triazole Schiff base derivative.
Guo-Wei Jiang et al.
The Journal of international medical research, 48(2), 300060520903874-300060520903874 (2020-02-12)
Determination of flavour components in natural vanilla extracts and synthetic flavourings by mixed micellar electrokinetic capillary chromatography.
Boyce, et al.
Analytica Chimica Acta, 485(2), 179-186 (2003)
Determination of vanillin and related flavor compounds in natural vanilla extracts and vanilla-flavored foods by thin layer chromatography and automated multiple development.
Belay MT and Poole CF.
Chromatographia, 37(7-8), 365-373 (1993)
Astragaloside IV promotes the eNOS/NO/cGMP pathway and improves left ventricular diastolic function in rats with metabolic syndrome.
Xin Lin et al.
The Journal of international medical research, 48(1), 300060519826848-300060519826848 (2019-03-08)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service