Skip to Content
Merck
All Photos(1)

Key Documents

90340

Sigma-Aldrich

Triethylamine

puriss. p.a., ≥99.5% (GC)

Synonym(s):

N,N-Diethylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
Beilstein:
605283
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.5 (vs air)

Quality Level

vapor pressure

51.75 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

593 °F

shelf life

36 mo.

expl. lim.

8 %

impurities

≤0.1% water

evapn. residue

≤0.003%

refractive index

n20/D 1.401 (lit.)
n20/D 1.401

bp

88.8 °C (lit.)

mp

−115 °C (lit.)

density

0.727 g/mL at 20 °C
0.726 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.2 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.02 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

functional group

amine

SMILES string

CCN(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethylamine is a tertiary amine. It has been reported as a promising precipitating reagent for phosphomolybdic acid. Et3N/formic acid system has been used for the asymmetric transfer hydrogenation (ATH) of methoxy-substituted derivatives.

Triethylamine is a strong base and precipitating agent widely used in Heck reaction and to prepare esters and amides from acyl chlorides.

Application

Triethylamine may be used in the following studies:
  • As a base for the Heck reactions.
  • Synthesis of spirocycles via Heck cyclization.
  • Preparation of triethylammonium picrate and 0.1M triethylamine hydrochloride.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Triethanolamine-Mediated Magnetically Separable Fe3O4- Pd Nanoparticles Catalyzed Heck Reaction under Ligand-Free Conditions
Kumar PC, et al.
ChemistrySelect, 6(31), 7944-7949}-7944-7949} (2021)
Asymmetric reduction of electron-rich ketones with tethered Ru(II)/TsDPEN catalysts using formic acid/triethylamine or aqueous sodium formate.
Soni R, et al.
The Journal of Organic Chemistry, 80(13), 6784-6793 (2015)
Zhao et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 6(5), 843-848 (2000-05-29)
The vinylation of iodobenzene with methyl acrylate has been studied with several supported palladium catalysts in N-methylpyrrolidone in the presence of triethylamine and/or sodium carbonate. The reaction can be performed in air without any solubilizing or activating ligands. It was
Biodegradable porous polymers through emulsion templating.
Lumelsky Y and Silverstein MS.
Macromolecules, 42(5), 1627-1633 (2009)
Acid-base reactions in non-dissociating solvents. Acetic acid and triethylamine in carbon tetrachloride and chloroform.
Barrow GM and Yerger EA.
Journal of the American Chemical Society, 76(20), 5211-5216 (1954)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service