Skip to Content
Merck
All Photos(1)

Key Documents

89273

Supelco

Borate buffer

suitable for HPLC, LiChropur, 0.127-0.140 M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161705
NACRES:
NA.21
Pricing and availability is not currently available.

form

liquid

Quality Level

quality

LiChropur

concentration

0.127-0.140 M

technique(s)

HPLC: suitable

refractive index

n20/D 1.330-1.340

pH

10.0-10.4

density

1.000-1.020 g/mL at 20 °C

UV absorption

λ: 210 nm Amax: ≤0.08
λ: 220 nm Amax: ≤0.03
λ: 230 nm Amax: ≤0.02
λ: 260 nm Amax: ≤0.01
λ: 500 nm Amax: ≤0.01

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. may i ask how to prepare 5% of borate buffer?Thanks

    1 answer
    1. The most accurate way to represent the concentration of Product No. 89273 is in molarity due to the multiple borate species that may be present in the buffer. However, assuming sodium tetraborate decahydrate predominates, the mass percentage solution concentration (m/m) can be calculated using the molecular weight of sodium tetraborate decahydrate and the molar concentration/density of the solution reported in the lot-specific Certificate of Analysis.

      The specifications for solution molar concentration are between 0.127 M and 0.140 M, whereas the specifications for solution density are between 1.000 g/mL and 1.020 g/mL (at 20 °C). The maximum and minimum specified percentage solutions (m/m) can be calculated from these values. The mass percentage solution is expressed as the total mass of the solute divided by the total mass of the solution, multiplied by 100.

      Using the minimum specified molar concentration of 0.127 M as an example, this can be expressed as 0.127 moles per liter of solution or 0.127 moles per 1000 mL of solution. Multiplying this 0.127 moles per 1000 mL of solution by the molecular weight of the assumed sodium tetraborate decahydrate (381.37 g/mol), dividing it by the maximum specified density (1.020 g/mL), and multiplying the result by 100 yields a minimum percentage solution (m/m) of 4.75%.

      Repeating the calculation using the maximum specified molar concentration of 0.140 M and the minimum specified density of 1.000 g/mL yields a maximum percentage solution (m/m) of 5.34%. An exact percentage solution (m/m) would need to be calculated from the specific values listed in the lot-specific Certificate of Analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service