Skip to Content
Merck
All Photos(1)

Key Documents

74042

Millipore

Optochin Disks

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

product line

BioChemika

form

disc

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | susceptibility testing: suitable

application(s)

clinical testing
food and beverages

microbiology

storage temp.

2-8°C

suitability

Pneumococcus spp.
Streptococcus viridans

General description

The optochin test is a useful diagnostic aid for identification/differentiation of pneumococci and viridans streptococci. Optochin (ethyl hydrocuprein hydrochloride) is inhibitory for pneumococcal growth whereas other streptococci show good growth or a very small zone of inhibition. This test is performed for the diagnosis of pneumococcal infections.

Application

Optochin Disks has been used to identify pneumococci.

Packaging

Sterile filter paper discs (50pcs) impregnated with optochin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of Clinical Streptococcus pneumoniae Isolates among other Alpha and Nonhemolytic Streptococci by Use of the Vitek MS Matrix-Assisted Laser Desorption Ionization?Time of Flight Mass Spectrometry System
Damien Dubois
Journal of Clinical Microbiology, 51(6), 1861-1867 (2013)
Biofilm Formation Enhances Fomite Survival of Streptococcus pneumoniae and Streptococcus pyogenes
Laura R. Marks
Infection and Immunity, 1141-1146 (2013)

Articles

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service