Skip to Content
Merck
All Photos(1)

Key Documents

73483

Supelco

Dopamine-1,1,2,2-d4 hydrochloride

analytical standard

Synonym(s):

2-(3,4-Dihydroxyphenyl)ethyl-1,1,2,2-d4-amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8D4H7O2N.HCl
CAS Number:
Molecular Weight:
193.66
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥96.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

248-250 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical
veterinary

format

neat

mass shift

M+4

storage temp.

−20°C

SMILES string

Cl.[2H]C([2H])(N)C([2H])([2H])c1ccc(O)c(O)c1

InChI

1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H/i3D2,4D2;

InChI key

CTENFNNZBMHDDG-URZLSVTISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dopamine-1,1,2,2-d4 hydrochloride may be used as an analytical standard for the analysis of dopamine in biological samples using on-line solid phase extraction method with high performance liquid chromatography-tandem mass spectrometry (XLC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

LC-MS identity conforms

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Automated mass spectrometric analysis of urinary free catecholamines using on-line solid phase extraction.
De Jong WH, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 878(19), 1506- 1512 (2010)
Adriana Basnakova et al.
Neurobiology of stress, 15, 100403-100403 (2021-10-12)
The response of an animal to a sensory stimulus depends on the nature of the stimulus and on expectations, which are mediated by spontaneous activity. Here, we ask how circadian variation in the expectation of danger, and thus the response
Celia Herrera-Rincon et al.
NPJ Regenerative medicine, 5, 2-2 (2020-02-13)
Infections have numerous effects on the brain. However, possible roles of the brain in protecting against infection, and the developmental origin and role of brain signaling in immune response, are largely unknown. We exploited a unique Xenopus embryonic model to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service