Skip to Content
Merck
All Photos(3)

Key Documents

46453

Supelco

6-Methyl-2-thiouracil

VETRANAL®, analytical standard

Synonym(s):

4-Hydroxy-2-mercapto 6-methylpyrimidine, MZU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2OS
CAS Number:
Molecular Weight:
142.18
Beilstein:
115648
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

~330 °C (dec.) (lit.)

application(s)

clinical testing

format

neat

SMILES string

CC1=CC(=O)NC(=S)N1

InChI

1S/C5H6N2OS/c1-3-2-4(8)7-5(9)6-3/h2H,1H3,(H2,6,7,8,9)

InChI key

HWGBHCRJGXAGEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Krivánková et al.
Electrophoresis, 17(12), 1959-1963 (1996-12-01)
Capillary zone electrophoresis was optimized for the separation of thiouracil, methylthiouracil and propylthiouracil. Methylthiouracil could be determined in various types of urine (human, bovine, horse), either without any pretreatment or in ethyl acetate extracts, within 15 min. For identification, the
J Vanden Bussche et al.
Analytica chimica acta, 637(1-2), 2-12 (2009-03-17)
Thyreostatic drugs (TS), illegally administrated to livestock for fattening purposes, are banned in the European Union since 1981 (Council Directive 81/602/EC). This paper reviews the trends in the analytical approaches for the determination of TS drugs in biological matrices. After
Joseph F Contrera et al.
Regulatory toxicology and pharmacology : RTP, 38(3), 243-259 (2003-11-19)
MDL QSAR (formerly SciVision QSAR IS) software is one of the several software systems under evaluation by the Informatics and Computational Safety Analysis Staff (ICSAS) of the FDA Center for Drug Evaluation and Research for regulatory and scientific decision support
A Heeremans et al.
The Analyst, 123(12), 2629-2632 (1999-08-06)
In addition to methods for the determination of residues, there is an important need for knowledge of the fate and excretion of growth promoting substances in fattening animals. In court, often simple questions are asked, such as, over what period
Yue Wei et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 854(1-2), 239-244 (2007-05-25)
Based on the sensitizing effect of formaldehyde on the chemiluminescence (CL) reaction of propylthiouracil (PTU) and methylthiouracil (MTU) with acidic potassium permanganate and the combination technique of high-performance liquid chromatography (HPLC), a sensitive, selective and simple post-column CL detection method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service