Skip to Content
Merck
All Photos(1)

Key Documents

42637

Supelco

Nickel Standard for IC

TraceCERT®, 1000 mg/L Ni+2 in nitric acid (nominal concentration)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
₪1,813.00
25 MG
₪3,985.00
50 MG
₪4,172.00

₪1,813.00


Availability
Please contact Customer Service for Availability

To order products, please contact your local dealer.

Select a Size

Change View
10 MG
₪1,813.00
25 MG
₪3,985.00
50 MG
₪4,172.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

₪1,813.00


Availability
Please contact Customer Service for Availability

To order products, please contact your local dealer.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1000 mg/L Ni+2 in nitric acid (nominal concentration)

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
434929015767902
application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

format

single component solution

format

single component solution

format

single component solution

format

single component solution

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

concentration

1000 mg/L Ni+2 in nitric acid (nominal concentration)

concentration

1000 mg/L Na+ in water (nominal concentration)

concentration

1000 mg/L cyanide in water (nominal concentration)

concentration

1000 mg/L Zn+2 in nitric acid (nominal concentration)

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Linkage

Visit the IC Portal to learn more

Analysis Note

1000 mg/L Ni2+ in ≤0.1% nitric acid, prepared with high purity Ni metal, HNO3 and water

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service