Skip to Content
Merck
All Photos(2)

Documents

40430

Supelco

Potassium sorbate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Potassium 2,4-hexadienoate, Sorbic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOK
CAS Number:
Molecular Weight:
150.22
Beilstein:
5357554
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[K+].C\C=C\C=C\C([O-])=O

InChI

1S/C6H8O2.K/c1-2-3-4-5-6(7)8;/h2-5H,1H3,(H,7,8);/q;+1/p-1/b3-2+,5-4+;

InChI key

CHHHXKFHOYLYRE-STWYSWDKSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Introduction to Food Toxicology (2009)
Control of citrus postharvest green mold and sour rot by potassium sorbate combined with heat and fungicides.
Smilanick L J, et al.
Postharvest biology and technology, 47(2), 226-238 (2008)
A Comparison between UV Spectrophotometer and High-performance Liquid Chromatography Method for the Analysis of Sodium Benzoate and Potassium Sorbate in Food Products.
Mahboubifar M, et al.
Food Analytical Methods, 4(2), 150-154 (2011)
Jonathan Byrne et al.
Journal of pharmaceutical and biomedical analysis, 96, 111-117 (2014-04-16)
A novel stability-indicating reversed phase high performance liquid chromatographic (RP-HPLC) method for the simultaneous assay of betamethasone-17-valerate, fusidic acid and potassium sorbate as well as methyl- and propylparaben in a topical cream preparation has been developed. A 100mm×3.0mm ID. Ascentis
Ana Oliveira et al.
Food chemistry, 170, 74-83 (2014-10-13)
Strawberries purées are incorporated in foods and subjected to pH modulation according to the expected final food matrix. The effect of pH on strawberry polyphenols stored at 4 and 23 °C for 90 days was evaluated. Total antioxidant activity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service