Skip to Content
Merck
All Photos(1)

Key Documents

19358

Supelco

Methyl butyrate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOCH3
CAS Number:
Molecular Weight:
102.13
Beilstein:
1740743
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 30 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

3.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.385 (lit.)
n20/D 1.388

bp

102-103 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCC(=O)OC

InChI

1S/C5H10O2/c1-3-4-5(6)7-2/h3-4H2,1-2H3

InChI key

UUIQMZJEGPQKFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl butyrate is a clear colourless liquid which is slightly soluble in water. It adds flavour and odour to fruits and flowers. It has a characteristic pineapple odour.

Application

It was used as reference material in analysing FAME from different fat classes using proton transfer reaction mass spectrometry (PTR-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.0 °F

Flash Point(C)

11.67 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Authentication of feeding fats: classification of animal fats, fish oils and recycled cooking oils.
Van Ruth, S. M., et al.
Anim. Feed Sci. Technol., 155.1, 65-73 (2010)
S H Sterri et al.
Biochemical pharmacology, 34(15), 2779-2785 (1985-08-01)
A radiochemical assay for carboxylesterase based on the substrate methyl[1-14C]butyrate is described. The blank value corresponds to 1.04 micrograms (liver)-1.44 mg (plasma) of tissues with the highest and lowest activity respectively, which constitute the sensitivity of the method. The hydrolysis
D Roy et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 15(17), 3839-3847 (2014-09-10)
An easily dispersible multiwalled carbon nanotube (MWCNT) derivative is prepared, and provides a platform for the synthesis of the phenyl butyric acid methyl ester (PCBM) analog. The carbene addition reaction of MWCNTs makes derivatives that are less soluble in organic
C D Hull et al.
Journal of chemical ecology, 27(5), 889-906 (2001-07-27)
Single-unit electrophysiology was used to record the nerve impulses from the carbon dioxide receptors of female Queensland fruit flies, Bactroera tryoni. The receptors responded to stimulation in a phasic-tonic manner and also had a period of inhibition of the nerve
A Nakagawa et al.
Journal of biochemistry, 96(3), 815-820 (1984-09-01)
Purified Pseudomonas fluorescens lipase [EC 3.1.1.3] exhibited slight activity on water-soluble esters such as methyl butyrate, and this activity was increased on addition of Brij 58 (20 oxyethylene hexadecyl ether) to the solution. This stimulating effect of Brij 58 on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service