Skip to Content
Merck
All Photos(1)

Key Documents

16669

Sigma-Aldrich

Magenta-Gal

reagent for selection of recombinant bacterial clones

Synonym(s):

5-Bromo-6-chloro-3-indolyl-β-D-galactopyranoside, Mag-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Beilstein:
1552602
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for molecular biology

Quality Level

sterility

non-sterile

Assay

≥98.0% (HPLC)

form

powder

solubility

DMF: soluble
DMSO: soluble

suitability

suitable for β-galactosidase test

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3cc(Cl)c(Br)cc23)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H15BrClNO6/c15-6-1-5-8(2-7(6)16)17-3-9(5)22-14-13(21)12(20)11(19)10(4-18)23-14/h1-3,10-14,17-21H,4H2/t10-,11-,12+,13-,14-/m1/s1

InChI key

CHRVKCMQIZYLNM-RKQHYHRCSA-N

General description

Magenta-Gal is a chromogenic substrate for β-galactosidase, used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Magenta-Gal is designed to replace X-Gal in blue-white selection of recombinant bacterial colonies with the lac+ phenotype.

Application

Suitable for use in selection of recombinant bacterial colonies with the lac+ phenotype. Also suitable for use immunoblotting and immunocytochemical assays.

Principle

When Magenta-Gal is hydrolyzed by β-galactosidase, the resulting product will form a red precipitate. Lac+ colonies grown in the presence of Magenta-Gal turn an intense red color, allowing for easy differentiation between lac+ and lac- colonies.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeon-Jin Kim et al.
Developmental biology, 397(1), 129-139 (2014-12-03)
Members of the fibroblast growth factor (FGF) family play important roles during various developmental processes including eye development. FRS (FGF receptor substrate) proteins bind to FGFR and serve as adapters for coordinated assembly of multi-protein complexes involved in Ras/MAPK and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service