Skip to Content
Merck
All Photos(1)

Key Documents

16025

Millipore

Brilliant Green Bile Lactose Broth

NutriSelect® Basic, suitable for microbiology

Synonym(s):

BRILA-Broth, Brilliant Green Bile Bouillon 2%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85
Pricing and availability is not currently available.

Agency

according to GB 4789.3-2016
according to ISO 4831:2006
according to ISO 4832:2006

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

brilliant green, 0.0133 g/L
lactose, 10 g/L
ox-bile (dried), 20 g/L
peptone, 10 g/L

manufacturer/tradename

NutriSelect® Basic

final pH

7.2±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages
water monitoring

microbiology

suitability

selective for Enterobacter spp.
selective for Escherichia coli
selective for coliforms

Application

For the selective enrichment and enumeration by titer determination or MPN method of E. coli and other fecal coliform from water, milk, food and other material.

Preparation Note

Dissolve 40 g in 1 litre distilled water. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new synthetic, selective, liquid medium for determination of coliform bacteria.
G Szita et al.
Acta veterinaria Hungarica, 34(3-4), 145-149 (1986-01-01)

Articles

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Protocols

General guidance for the detection and enumeration of Enterobacteriaceae in food according to EN-ISO 8523:1991 and EN-ISO 4832:1991, respectively.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service