Skip to Content
Merck
All Photos(1)

Key Documents

14690

Supelco

Bis(cyclohexanone)oxaldihydrazone

for spectrophotometric det. of Cu, ≥99.0%

Synonym(s):

Cuprizon, Cuprizone, Oxalic acid bis(cyclohexylidenehydrazide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N4O2
CAS Number:
Molecular Weight:
278.35
Beilstein:
2388004
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.0% (N)
≥99.0%

form

powder

quality

for spectrophotometric det. of Cu

technique(s)

UV/Vis spectroscopy: suitable

mp

210-214 °C (lit.)
210-214 °C

SMILES string

O=C(N\N=C1\CCCCC1)C(=O)N\N=C2/CCCCC2

InChI

1S/C14H22N4O2/c19-13(17-15-11-7-3-1-4-8-11)14(20)18-16-12-9-5-2-6-10-12/h1-10H2,(H,17,19)(H,18,20)

InChI key

DSRJIHMZAQEUJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cuprizone (Bis(cyclohexanone)oxaldihydrazone) is a physically unstable compound and is a well-known neurotoxic agent.

Application

Cuprizone (Bis(cyclohexanone)oxaldihydrazone) was used for toxic demyelination in the central nervous system in order to investigate the pathobiology of remyelination in the corpus callosum, so as to study the human demyelinating diseases such as multiple sclerosis. It was also used a model to study components of neuroinflammation.

Other Notes

For the spectrophotometric determination of Cu(II)
In the presence of a copper salt, this ligand catalyzes Ullmann-type C-N bond forming reactions under mild, aqueous conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhu, X., et al.
Synthesis, 23, 3955-3962 (2006)
Cuprizone treatment induces distinct demyelination, astrocytosis, and microglia cell invasion or proliferation in the mouse cerebellum.
Groebe A
Cerebellum, 8 (3), 163-174 (2009)
R.K. Rohde
Analytical Chemistry, 38, 911-911 (1966)
The protective role of nitric oxide in a neurotoxicant-induced demyelinating model.
Arnett H
Journal of Immunology, 168 (1), 427-433 (2002)
Cortical demyelination is prominent in the murine cuprizone model and is strain-dependent.
Thomas S
The American Journal of Pathology, 174 (4), 1053-1061 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service