Skip to Content
Merck
All Photos(1)

Key Documents

05393

Supelco

Eukitt® UV

for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41191506

grade

for microscopy

Quality Level

form

liquid

refractive index

n20/D 1.480-1.490

viscosity

350-550 mPa.s

suitability

in accordance for application
suitable for microscopy

General description

EUKITT® UV is a UV-curing mounting medium with all the well-known beneficial properties of the EUKITT® series and a true innovation in the mounting technology. EUKITT® UV does not contain any solvents, therefore only 50% of the usual amount is needed compared to conventional systems. Using EUKITT® UV almost no shrinkage can be detected.
The mounting medium may be used on tissue sections in non-polar solvents such as xylene or xylene substitutes as well as directly on sections in pure alcohols.
The viscosity of EUKITT® UV is very user-friendly. After being applied to the tissue, the cover glass can be placed as usual. Subsequently, a transfer break of at least 120 sec. is maintained prior to irradiation in order to ensure an ideal penetration into the tissue.

Features and Benefits

  • Curing time: 10 – 30 seconds
  • No yellowing after long storage
  • No brittle fracture
  • Suitable for all tissue types

Legal Information

Eukitt is a registered trademark of Orsatec GmbH

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service