Skip to Content
Merck
All Photos(1)

Key Documents

28-5940

Sigma-Aldrich

Sulfuric acid solution

0.5 M

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

reagent

form

liquid

availability

available only in Japan

concentration

0.5 M
1 N

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

João Almeida et al.
Nature, 502(7471), 359-363 (2013-10-08)
Nucleation of aerosol particles from trace atmospheric vapours is thought to provide up to half of global cloud condensation nuclei. Aerosols can cause a net cooling of climate by scattering sunlight and by leading to smaller but more numerous cloud
Ying Gu et al.
Cell research, 25(5), 604-620 (2015-03-21)
Hepatitis E virus (HEV), a non-enveloped, positive-sense, single-stranded RNA virus, is a major cause of enteric hepatitis. Classified into the family Hepeviridae, HEV comprises four genotypes (genotypes 1-4), which belong to a single serotype. We describe a monoclonal antibody (mAb)
Francesco Riccobono et al.
Science (New York, N.Y.), 344(6185), 717-721 (2014-05-17)
Atmospheric new-particle formation affects climate and is one of the least understood atmospheric aerosol processes. The complexity and variability of the atmosphere has hindered elucidation of the fundamental mechanism of new-particle formation from gaseous precursors. We show, in experiments performed
Andreas Recke et al.
Journal of immunology (Baltimore, Md. : 1950), 193(4), 1600-1608 (2014-07-16)
Subepidermal autoimmune blistering dermatoses (AIBD) are prototypic autoantibody-mediated diseases. In epidermolysis bullosa acquisita (EBA), an autoimmune disease with severe and chronic skin blistering, autoantibodies are directed against type VII collagen. IgG is the predominant autoantibody isotype of EBA, the pathogenicity
Jolan E Walter et al.
The Journal of clinical investigation, 125(11), 4135-4148 (2015-10-13)
Patients with mutations of the recombination-activating genes (RAG) present with diverse clinical phenotypes, including severe combined immune deficiency (SCID), autoimmunity, and inflammation. However, the incidence and extent of immune dysregulation in RAG-dependent immunodeficiency have not been studied in detail. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service