Skip to Content
Merck
All Photos(1)

Key Documents

28-5250

Sigma-Aldrich

Strontium chloride hexahydrate

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrCl2 · 6H2O
CAS Number:
Molecular Weight:
266.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥98.0%
grade:
SAJ first grade
form:
solid

grade

SAJ first grade

Assay

≥98.0%

form

solid

availability

available only in Japan

mp

115 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Sr]Cl

InChI

1S/2ClH.6H2O.Sr/h2*1H;6*1H2;/q;;;;;;;;+2/p-2

InChI key

AMGRXJSJSONEEG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Li et al.
American journal of dentistry, 25(4), 210-214 (2012-10-23)
To investigate the laboratory changes in dentin tubule occlusion morphology during short term use of desensitizing products as evaluated by electron microscopy and an image analysis. Freshly extracted human third molar teeth were collected at random and 40 dentin discs
Serdal Kirmizialtin et al.
Biophysical journal, 102(4), 829-838 (2012-03-06)
The distributions of different cations around A-RNA are computed by Poisson-Boltzmann (PB) equation and replica exchange molecular dynamics (MD). Both the nonlinear PB and size-modified PB theories are considered. The number of ions bound to A-RNA, which can be measured
Diane Cummins
American journal of dentistry, 23 Spec No A, 3A-13A (2011-02-03)
To provide a brief overview of the diagnosis, epidemiology, etiology and clinical management of dentin hypersensitivity, to discuss technical approaches to relieve sensitivity, with special emphasis on dentin tubule occlusion and the clinical evidence for efficacy of desensitizing toothpastes based
Karen A Autio et al.
Current treatment options in oncology, 13(2), 174-188 (2012-04-25)
Skeletal metastases threaten quality of life, functionality, and longevity in patients with metastatic castration-resistant prostate cancer (mCRPC). Therapeutic strategies for bone metastases in prostate cancer can palliate pain, delay/prevent skeletal complications, and prolong survival. Pharmacologic agents representing several drug classes
Salil Vengalil et al.
Current opinion in supportive and palliative care, 6(3), 310-315 (2012-06-20)
Bone metastases in prostate cancer are often the cause of significant morbidity in patients with castrate-resistant disease, and several studies have shown significant pain palliation with systemic radionuclide treatment. The purpose of this review is to discuss the place of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service