Skip to Content
Merck
All Photos(1)

Key Documents

28-2590

Sigma-Aldrich

Sodium diethyldithiocarbamate trihydrate

JIS special grade, ≥92.0%

Synonym(s):

Cupral, Diethyldithiocarbamic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCSSNa · 3H2O
CAS Number:
Molecular Weight:
225.31
Beilstein:
3920507
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥92.0%
grade:
JIS special grade
form:
solid

grade

JIS special grade

vapor density

5.9 (vs air)

Assay

≥92.0%

form

solid

availability

available only in Japan

mp

95-98.5 °C (lit.)

storage temp.

15-25°C

SMILES string

[H]O[H].[H]O[H].[H]O[H].CCN(CC)C(=S)S[Na]

InChI

1S/C5H11NS2.Na.3H2O/c1-3-6(4-2)5(7)8;;;;/h3-4H2,1-2H3,(H,7,8);;3*1H2/q;+1;;;/p-1

InChI key

WWGXHTXOZKVJDN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues.
Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO2 has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

Biochem/physiol Actions

Inhibits induction of macrophage nitric oxide synthase.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nini Skovgaard et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 303(5), R487-R494 (2012-06-29)
Hypoxic pulmonary vasoconstriction (HPV) is an adaptive response that diverts pulmonary blood flow from poorly ventilated and hypoxic areas of the lung to more well-ventilated parts. This response is important for the local matching of blood perfusion to ventilation and
Zdenek Skrott et al.
Mini reviews in medicinal chemistry, 12(12), 1184-1192 (2012-08-31)
The idea of "repurposing" of existing drugs provides an effective way to develop and identify new therapies. Disulfiram (Antabuse), a drug commonly used for the treatment of alcoholism, shows promising anticancer activity in both preclinical and clinical studies. In the
A C Costa et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 105, 251-258 (2013-01-15)
Theoretical and experimental bands have been assigned for the Fourier Transform Infrared (FT-IR) and Fourier Transform Raman (FT-Raman) spectra of the bis-(diethydithiocarbamate)Zn(II) complex, [Zn(DDTC)(2)]. The calculations and spectra interpretation have been based on the DFT/B3LYP method, infrared and Raman second
Bhupesh Sharma et al.
Progress in neuro-psychopharmacology & biological psychiatry, 38(2), 207-215 (2012-04-19)
Dementia of vascular origin or vascular dementia (VaD) is considered as the second commonest form of dementia after Alzheimer's disease (AD). In the last ten years various researchers have reported a strong association of hyperhomocysteinemia (HHcy), hyperlipidemia (HL) and dementia.
Kuno Lehmann et al.
Annals of surgery, 256(5), 730-737 (2012-10-26)
The aim of hyperthermic intraperitoneal chemotherapy (HIPEC) is to eradicate microscopic residual tumor after radical surgical tumor excision in patients with peritoneal carcinomatosis. The common use of antineoplastic agents such as mitomycin C, doxorubicin, or oxaliplatin with hyperthermia fails to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service