Skip to Content
Merck
All Photos(1)

Key Documents

24-1180

Sigma-Aldrich

Petroleum ether

JIS special grade, ≥90.0%

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
grade:
JIS special grade
bp:
30-60 °C (lit.)
vapor pressure:
25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

grade

JIS special grade

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

Assay

≥90.0%

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)

density

0.64 g/mL at 25 °C (lit.)

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sujith Prasad W Kumarasinghe et al.
International journal of dermatology, 41(12), 877-880 (2002-12-21)
Many methods have been employed, with variable success, in the treatment of cutaneous myiasis caused by Chrysomya species. Experiment 1: to assess the larvicidal effect of mineral turpentine (MT) and the main ingredient of MT, low aromatic white spirits (LAWS)
Catherine F Poh et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(22), 6716-6722 (2006-11-24)
Genetically altered cells could become widespread across the epithelium of patients with oral cancer, often in clinically and histologically normal tissue, and contribute to recurrent disease. Molecular approaches have begun to yield information on cancer/risk fields; tissue optics could further
S C Bondy et al.
Archives of toxicology, 69(6), 410-414 (1995-01-01)
Levels of glutathione and activity of glutamine synthetase were assayed in organs of rats following inhalation of a heterogeneous solvent mixture containing both aliphatic and aromatic hydrocarbons. This mixture was administered for 3 weeks (6 h daily) at two levels
I P Dick et al.
Human & experimental toxicology, 16(11), 652-657 (1998-01-14)
1. The absorption of lindane through human skin was assessed in vitro using static diffusion cells with 50% aqueous ethanol as the receptor fluid. Four formulations of lindane were applied, three of which were commercial preparations. The dermal distribution of
A Steensgaard et al.
Neurotoxicology, 17(3-4), 785-792 (1996-10-01)
Several epidemiological studies of workers occupationally exposed to white spirit show that neuropsychiatric disorders are a frequent cause of early disability pension in this population compared with non-exposed controls. In the rat, we have demonstrated that exposure to different kinds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service