Skip to Content
Merck
All Photos(1)

Documents

19-1080

Sigma-Aldrich

D-Mannitol

SAJ first grade, ≥99.0%

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
EC Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.0%

availability

available only in Japan

mp

167-170 °C (lit.)

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Badal Tejedor et al.
International journal of pharmaceutics, 486(1-2), 315-323 (2015-04-07)
Tablets are the most convenient form for drug administration. However, despite the ease of manufacturing problems such as powder adhesion occur during the production process. This study presents surface and structural characterization of tablets formulated with commonly used excipients (microcrystalline
Heikki O Koskela et al.
Chest, 124(4), 1341-1349 (2003-10-14)
To compare the sensitivity and validity of mannitol, histamine, and cold air challenges to demonstrate the effect of inhaled corticosteroids (ICS) in asthma. A prospective study. Seventeen patients with recently diagnosed, steroid-naive asthma who fulfilled the diagnostic criteria of Finnish
Heikki O Koskela et al.
Chest, 125(6), 1985-1992 (2004-06-11)
To define whether coughing during mannitol challenge is a nonspecific side effect of this challenge or is associated with asthma. A prospective study. University hospital. Thirty-seven steroid-naive, asthmatic subjects and 10 healthy subjects. The participants completed a symptom questionnaire, recorded
Traumatic intracranial hypertension.
Nino Stocchetti et al.
The New England journal of medicine, 370(22), 2121-2130 (2014-05-30)
Diana Bilton et al.
Thorax, 69(12), 1073-1079 (2014-09-24)
Bronchiectasis is characterised by excessive production of mucus and pulmonary exacerbations. Inhaled osmotic agents may enhance mucociliary clearance, but few long-term clinical trials have been conducted. To determine the impact of inhaled mannitol on exacerbation rates in patients with non-cystic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service