Skip to Content
Merck
All Photos(1)

Key Documents

19-0280

Sigma-Aldrich

Magnesium hydroxide

SAJ first grade, ≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(OH)2
CAS Number:
Molecular Weight:
58.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥95.0%
grade:
SAJ first grade
form:
powder

grade

SAJ first grade

Assay

≥95.0%

form

powder

availability

available only in Japan

pH

9.5-10.5

mp

350 °C (lit.)

SMILES string

O[Mg]O

InChI

1S/Mg.2H2O/h;2*1H2/q+2;;/p-2

InChI key

VTHJTEIRLNZDEV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tengfei Lv et al.
Journal of hazardous materials, 237-238, 121-132 (2012-09-11)
The present study explores the potential of Mg-Ca-Al (NO(3)) hydrotalcite-like compounds (MgCaAlNO(3)-HTlcs) for the removal of fluoride from protein solutions. In this study, the Mg(3-x)Ca(x)AlNO(3)-HTlcs (x=0-3, x is the mol.% of Ca) were synthesized and characterized by SEM, XRD, FTIR
Anna L Harrison et al.
Environmental science & technology, 47(1), 126-134 (2012-07-10)
Atmospheric CO(2) is sequestered within ultramafic mine tailings via carbonation of Mg-bearing minerals. The rate of carbon sequestration at some mine sites appears to be limited by the rate of CO(2) supply. If carbonation of bulk tailings were accelerated, large
Umberto Costantino et al.
Recent patents on nanotechnology, 6(3), 218-230 (2012-07-04)
This review treats the recent patents and related literature, mainly from the Authors laboratories, on biomedical and food packaging applications of nano-composites constituted of biodegradable polymers filled with micro or nano crystals of organically modified Layered Double Hydroxides of Hydrotalcite
Xiaohong Pan et al.
ACS applied materials & interfaces, 5(3), 1137-1142 (2013-01-11)
Here we reported the antibacterial effect and related mechanism of three nano-Mg(OH)(2) slurries using Escherichia coli as model bacteria. X-ray diffraction (XRD), scanning electron microscopy (SEM) and laser particle size analysis revealed that the as-synthesized Mg(OH)(2)_(MgCl2), Mg(OH)(2)_(MgSO4) and Mg(OH)(2)_(MgO) are
Francys K V Moreira et al.
Carbohydrate polymers, 92(2), 1743-1751 (2013-02-13)
In this paper the mechanical reinforcement of nano-sized brucite, Mg(OH)(2) in a series of bionanocomposite films based on starch was investigated. Brucite nanoplates with an aspect ratio of 9.25 were synthesized by wet precipitation and incorporated into starch matrices at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service