Skip to Content
Merck
All Photos(1)

Key Documents

09-2460

Sigma-Aldrich

Tetraethyl orthosilicate

SAJ first grade, ≥95.0%

Synonym(s):

Orthosilicic acid tetraethyl ester, TEOS, Tetraethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si(OC2H5)4
CAS Number:
Molecular Weight:
208.33
Beilstein:
1422225
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

vapor density

7.2 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥95.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.382 (lit.)

bp

168 °C (lit.)

density

0.933 g/mL at 20 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)OCC

InChI

1S/C8H20O4Si/c1-5-9-13(10-6-2,11-7-3)12-8-4/h5-8H2,1-4H3

InChI key

BOTDANWDWHJENH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Commonly used as a precursor to prepare xerogel
Will interact with dodecylamine in the formation of intercalation compounds of H+-magadiite and used in a study of mixed-metal bioactive glasses.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seong Cheol Hong et al.
International journal of nanomedicine, 6, 3219-3231 (2012-01-13)
Superparamagnetic iron oxide nanoparticles (SPIONs) have been widely utilized for the diagnosis and therapy of specific diseases, as magnetic resonance imaging (MRI) contrast agents and drug-delivery carriers, due to their easy transportation to targeted areas by an external magnetic field.
Nils R Blumenthal et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(45), 16124-16129 (2014-10-29)
Extracellular soluble signals are known to play a critical role in maintaining neuronal function and homeostasis in the CNS. However, the CNS is also composed of extracellular matrix macromolecules and glia support cells, and the contribution of the physical attributes
Lin Lin et al.
Colloids and surfaces. B, Biointerfaces, 101, 97-100 (2012-07-17)
Hollow silica spheres with round mesoporous shells were synthesized by core-shell template method, using monodispersed cationic polystyrene particles as core, and TEOS (tetraethoxysilane) as the silica source to form shell. After calcination at 550°C, uniform spheres with a thin shell
Tom Hasell et al.
Advanced materials (Deerfield Beach, Fla.), 24(42), 5732-5737 (2012-08-30)
Macroscopic inorganic porous beads are imbibed with a "porous molecular additive" by simple solution processing techniques, resulting in controllable loading and increased surface area. The porous additive consists of soluble organic cage molecules that precipitate as microporous crystals when solutions
Evan M Hetrick et al.
Biomaterials, 30(14), 2782-2789 (2009-02-24)
The ability of nitric oxide (NO)-releasing silica nanoparticles to kill biofilm-based microbial cells is reported. Biofilms of Pseudomonas aeruginosa, Escherichia coli, Staphylococcus aureus, Staphylococcus epidermidis, and Candida albicans were formed in vitro and exposed to NO-releasing silica nanoparticles. Replicative viability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service