Skip to Content
Merck
All Photos(1)

Key Documents

09-0615

Sigma-Aldrich

Ethidium bromide

≥95.0%

Synonym(s):

3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, EtBr, Homidium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20BrN3
CAS Number:
Molecular Weight:
394.31
Beilstein:
3642536
EC Number:
MDL number:
UNSPSC Code:
12161505
PubChem Substance ID:

Assay

≥95.0%

availability

available only in Japan

mp

260-262 °C (dec.) (lit.)

SMILES string

[Br-].CC[n+]1c(-c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14

InChI

1S/C21H19N3.BrH/c1-2-24-20-13-16(23)9-11-18(20)17-10-8-15(22)12-19(17)21(24)14-6-4-3-5-7-14;/h3-13,23H,2,22H2,1H3;1H

InChI key

ZMMJGEGLRURXTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethidium bromide (EtBr) is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis. The fluorescence of EtBr increases 21-fold upon binding to double-stranded RNA and 25-fold on binding double-stranded DNA so that destaining the background is not necessary with a low stain concentration (10 μg/ml). Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids. It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA. Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase.

Frameshift mutagen which intercalates double-stranded DNA and RNA.

Biochem/physiol Actions

Ethidium bromide intercalates double-stranded DNA and RNA and acts as a frameshift mutagen. It can also be used in conjunction with acridine orange to differentiate between viable, apoptotic and necrotic cells.

Suitability

for biological purposes

Reconstitution

For staining a gel after electrophoresis, dilute a sample of the stock solution to 0.5 μg/ml with water and incubate the gel for 15-30 min. Destaining is usually not needed but can be carried out in water for 15 min if decreased background is necessary. The DNA bands can then be detected on a UV light box (254 nm wavelength). Ethidium bromide can also be incorporated into the gel and running buffer at 0.5 μg/ml and visualized immediately after electrophoresis.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Muta. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bobo W Mok et al.
PloS one, 3(4), e1982-e1982 (2008-04-24)
Antigenic variation is a subtle process of fundamental importance to the survival of a microbial pathogen. In Plasmodium falciparum malaria, PfEMP1 is the major variable antigen and adhesin expressed at the surface of the infected erythrocyte, which is encoded for
Wenfang Peng et al.
Nucleic acids research, 43(1), 406-417 (2014-12-17)
CRISPR-Cas systems provide a small RNA-based mechanism to defend against invasive genetic elements in archaea and bacteria. To investigate the in vivo mechanism of RNA interference by two type III-B systems (Cmr-α and Cmr-β) in Sulfolobus islandicus, a genetic assay
Ulrich Braunschweig et al.
Genome research, 24(11), 1774-1786 (2014-09-27)
Alternative splicing (AS) of precursor RNAs is responsible for greatly expanding the regulatory and functional capacity of eukaryotic genomes. Of the different classes of AS, intron retention (IR) is the least well understood. In plants and unicellular eukaryotes, IR is
Patrick Seitz et al.
PLoS genetics, 10(1), e1004066-e1004066 (2014-01-07)
The DNA uptake of naturally competent bacteria has been attributed to the action of DNA uptake machineries resembling type IV pilus complexes. However, the protein(s) for pulling the DNA across the outer membrane of Gram-negative bacteria remain speculative. Here we
Alexandra C I Depelsenaire et al.
The Journal of investigative dermatology, 134(9), 2361-2370 (2014-04-10)
Vaccines delivered to the skin by microneedles-with and without adjuvants-have increased immunogenicity with lower doses than standard vaccine delivery techniques such as intramuscular or intradermal injection. However, the mechanisms underlying this skin-mediated "adjuvant" effect are not clear. Here, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service