Skip to Content
Merck
All Photos(1)

Key Documents

07-0055

Sigma-Aldrich

Decahydronaphthalene, mixture of cis + trans

SAJ first grade, ≥98.0%

Synonym(s):

Decalin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein:
878165
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

SAJ first grade

vapor density

4.76 (vs air)

vapor pressure

42 mmHg ( 92 °C)
741 mmHg ( 188 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

482 °F

expl. lim.

0.7-4.9 %, 100 °F

availability

available only in Japan

refractive index

n20/D 1.474 (lit.)

bp

189-191 °C (lit.)

mp

−125 °C (lit.)

density

0.896 g/mL at 25 °C (lit.)

SMILES string

C1CCC2CCCCC2C1

InChI

1S/C10H18/c1-2-6-10-8-4-3-7-9(10)5-1/h9-10H,1-8H2

InChI key

NNBZCPXTIHJBJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Decalin is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken K Qian et al.
Journal of pharmaceutical sciences, 100(7), 2801-2815 (2011-02-22)
Spontaneous crystalline-to-amorphous phase transformation of organic or medicinal molecules in the presence of mesoporous materials has been observed, for which pathway was suggested to be via the vapor phase, that is, sublimation of the crystalline molecules followed by adsorption on
Mohit Kumar et al.
Nanoscale, 3(5), 2130-2133 (2011-03-30)
Naphthalene diimide (NDI) bolaamphiphilic molecules (1) self-assemble in water to form organic nanoparticles, which exhibit self-assembly induced preassociated excimer formation and hence an enhanced green fluorescence.
Sylvie Goncalves et al.
The Journal of organic chemistry, 76(9), 3274-3285 (2011-03-31)
An unprecedented and highly diastereoselective 6-endo-trig cyclization of 2-alkenyl-1,3-dithiolanes has been developed yielding trans-decalins, an important scaffold present in numerous di- and triterpenes. The novelty of this 6-endo-trig cyclization stands in the stepwise mechanism involving 2-alkenyl-1,3-dithiolane, acting as a novel
Lin Zhu et al.
Journal of natural products, 75(4), 567-571 (2012-03-08)
Five new decalin derivatives (1-5), together with two known compounds (6 and 7), were isolated from the ethyl acetate extract of red yeast rice. Their structures were elucidated by means of NMR and mass spectroscopic analyses. Monascusic lactone A (1)
Joseph B Lim et al.
The journal of physical chemistry. B, 116(1), 203-210 (2011-12-06)
A modification of the CHARMM36 lipid force field (C36) for cholesterol, henceforth, called C36c, is reported. A fused ring compound, decalin, was used to model the steroid section of cholesterol. For decalin, C36 inaccurately predicts the heat of vaporization (~10

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service