Skip to Content
Merck
All Photos(1)

Key Documents

05-7470

Sigma-Aldrich

L-Cysteine hydrochloride monohydrate

JIS special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOH · HCl · H2O
CAS Number:
Molecular Weight:
175.63
Beilstein:
5158059
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

solubility

water: 100 g/L at 25 °C

application(s)

peptide synthesis

SMILES string

O.Cl.N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2/t2-;;/m0../s1

InChI key

QIJRTFXNRTXDIP-JIZZDEOASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

NMDA glutamatergic receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerdt Müller et al.
Biotechnology for biofuels, 8, 187-187 (2015-11-27)
The emerging bioeconomy depends on improved methods for processing of lignocellulosic biomass to fuels and chemicals. Saccharification of lignocellulose to fermentable sugars is a key step in this regard where enzymatic catalysis plays an important role and is a major
Qinjian Zhao et al.
Human vaccines & immunotherapeutics, 10(3), 734-739 (2013-12-05)
Cryo-transmission electron microscopy (cryoTEM) is a powerful characterization method for assessing the structural properties of biopharmaceutical nanoparticles, including Virus Like Particle-based vaccines. We demonstrate the method using the Human Papilloma Virus (HPV) VLPs in GARDASIL®. CryoTEM, coupled to automated data
Edward T Chouchani et al.
Nature medicine, 19(6), 753-759 (2013-05-28)
Oxidative damage from elevated production of reactive oxygen species (ROS) contributes to ischemia-reperfusion injury in myocardial infarction and stroke. The mechanism by which the increase in ROS occurs is not known, and it is unclear how this increase can be
Feng Wang et al.
Cell, 153(6), 1379-1393 (2013-06-12)
Some species mount a robust antibody response despite having limited genome-encoded combinatorial diversity potential. Cows are unusual in having exceptionally long CDR H3 loops and few V regions, but the mechanism for creating diversity is not understood. Deep sequencing reveals
Tal Ilani et al.
Science (New York, N.Y.), 341(6141), 74-76 (2013-05-25)
Disulfide bond formation in secretory proteins occurs primarily in the endoplasmic reticulum (ER), where multiple enzyme families catalyze cysteine cross-linking. Quiescin sulfhydryl oxidase 1 (QSOX1) is an atypical disulfide catalyst, localized to the Golgi apparatus or secreted from cells. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service