Skip to Content
Merck
All Photos(1)

Key Documents

05-2810

Sigma-Aldrich

Chloroacetic acid

JIS special grade, ≥99.0%

Synonym(s):

Monochloroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COOH
CAS Number:
Molecular Weight:
94.50
Beilstein:
605438
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

JIS special grade

vapor density

3.26 (vs air)

vapor pressure

0.75 mmHg ( 20 °C)

Assay

≥99.0%

form

crystalline

availability

available only in Japan

bp

189 °C (lit.)

mp

60-63 °C (lit.)

solubility

water: soluble 3170 g/L at 10 °C

SMILES string

OC(=O)CCl

InChI

1S/C2H3ClO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

FOCAUTSVDIKZOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hany El-Hamshary et al.
International journal of biological macromolecules, 72, 1466-1472 (2014-12-03)
Poly(N-vinyl imidazole) (PVI) has been grafted onto carboxymethyl starch (CMS) in aqueous solution using potassium persulfate (KPS) as initiator. Reaction parameters that affect grafting efficiency and percentage grafting such as monomer and initiator concentration, the reaction temperature and time were
Maike M Schmidt et al.
Neurotoxicity research, 19(4), 628-637 (2010-07-16)
The chlorinated acetates monochloroacetate (MCA), dichloroacetate (DCA), and trichloroacetate (TCA) are generated in water disinfection processes and are formed during metabolic detoxification of industrial solvents such as trichloroethylene. In order to test for consequences of an exposure of brain cells
Brendon Y Chua et al.
Molecular pharmaceutics, 9(1), 81-90 (2011-12-14)
It has become increasingly recognized that polymer particle size can have a profound effect on the interactions of particle-based vaccines with antigen presenting cells (APCs) thereby influencing and modulating ensuing immune responses. With the aim of developing chitosan particle-based immunocontraceptive
Xuchun Li et al.
Environmental science & technology, 46(13), 7342-7349 (2012-06-12)
Most halogenated organic compounds (HOCs) are toxic and persistent, and their efficient destruction is currently a challenge. Here, we proposed a sulfite/UV (253.7 nm) process to eliminate HOCs. Monochloroacetic acid (MCAA) was selected as the target compound and was degraded
Wang Yanli et al.
Carbohydrate research, 344(13), 1764-1769 (2009-08-12)
Carboxymethyl starch (CMS) was obtained as a product of the reaction of starch and monochloroacetic acid (MCA) in the presence of sodium hydroxide. The influence of the molar ratio of NaOH/AGU, the molar ratio of MCA/AGU, the reaction time, reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service