Skip to Content
Merck
All Photos(1)

Documents

05-0390

Sigma-Aldrich

Calcium

SAJ first grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ca
CAS Number:
Molecular Weight:
40.08
Beilstein:
4241647
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.5%

form

solid

reaction suitability

reagent type: reductant

availability

available only in Japan

resistivity

3.5 μΩ-cm, 20°C

bp

1484 °C (lit.)

mp

850 °C (lit.)

density

1.54 g/mL at 25 °C (lit.)

SMILES string

[Ca]

InChI

1S/Ca

InChI key

OYPRJOBELJOOCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to Ca8Al3 and Ca13Al14; intermetallics with new metallic Zintl phases.7

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleni Gourni et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(10), 1719-1725 (2014-08-26)
Gastrin-releasing peptide receptors (GRPrs) are overexpressed on a variety of human cancers, providing the opportunity for peptide receptor targeting via radiolabeled bombesin-based peptides. As part of our ongoing investigations into the development of improved GRPr antagonists, this study aimed at
Xinyu Zhou et al.
Circulation research, 114(4), 706-716 (2014-02-15)
Trimeric intracellular cation channels (TRIC) represents a novel class of trimeric intracellular cation channels. Two TRIC isoforms have been identified in both the human and the mouse genomes: TRIC-A, a subtype predominantly expressed in the sarcoplasmic reticulum (SR) of muscle
Jun Suzuki et al.
Science (New York, N.Y.), 341(6144), 403-406 (2013-07-13)
A classic feature of apoptotic cells is the cell-surface exposure of phosphatidylserine (PtdSer) as an "eat me" signal for engulfment. We show that the Xk-family protein Xkr8 mediates PtdSer exposure in response to apoptotic stimuli. Mouse Xkr8(-/-) cells or human
Tsai-Wen Chen et al.
Nature, 499(7458), 295-300 (2013-07-23)
Fluorescent calcium sensors are widely used to image neural activity. Using structure-based mutagenesis and neuron-based screening, we developed a family of ultrasensitive protein calcium sensors (GCaMP6) that outperformed other sensors in cultured neurons and in zebrafish, flies and mice in
S Boonen et al.
Journal of internal medicine, 259(6), 539-552 (2006-05-18)
Combined calcium and vitamin D supplementation is an essential component of the management of osteoporosis, supported by a strong scientific rationale. The types of individuals who should receive calcium and vitamin D supplements are those: (i) patients with documented osteoporosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service