Skip to Content
Merck
All Photos(1)

Key Documents

NAD98-RO

Roche

NAD

Grade II, free acid

Synonym(s):

β-Nicotinamide adenine dinucleotide hydrate, β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2 · xH2O
CAS Number:
Molecular Weight:
663.43 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98% (dry weight)

form

lyophilized

mol wt

Mr 663.4

packaging

pkg of 1 g (10127981001)
pkg of 10 g (10621650001)
pkg of 5 g (10127990001)

manufacturer/tradename

Roche

color

off-white

pH

~3.0 (50 mg/mL in water)

λmax

260 nm

shipped in

wet ice

storage temp.

2-8°C

SMILES string

NC1=NC=NC2=C1N=CN2.O[C@@H]3[C@@H](COP(O)(OP(OC[C@@H](O4)[C@@H](O)[C@@H](O)[C@H]4[N+]5=CC=CC(C(N)=O)=C5)([O-])=O)=O)OC[C@@H]3O

InChI

1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)/t10-,11-,13-,14-,15-,16-,20-,21-/m1/s1

InChI key

BAWFJGJZGIEFAR-NNYOXOHSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinamide adenine dinucleotide (NAD) is a cofactor containing an adenylic acid and a nicotinamide-5′-ribonucleotide group linked together by a pyrophosphate moiety. It is ubiquitously found. In NAD+ complexes, the enzyme-cofactor interactions are highly conserved.[1]

Application

NAD has been used in boiled-blood agar plate as a supplement for the growth of Actinobacillus pleuropneumoniae colony.[2]

Biochem/physiol Actions

Electron acceptor
Nicotinamide adenine dinucleotide (NAD) acts as an electron carrier[3] and significantly participates in enzyme-catalyzed oxido-reduction processes[1] and many genetic processes.[4] NAD cycles between the oxidized (NAD+) and reduced (NADH) forms to maintain a redox balance necessary for continued cell growth.[4] This cofactor is also involved in microbial catabolism.[4] β-NAD acts as a substrate for various enzymes in several cellular processes.[5]

Packaging

Packaged by solid weight.

Quality

Absence of contaminants: LDH inhibitors are not detectable.
Organic solvents: <0.5% (GC)

Preparation Note

Storage conditions (working solution): An aqueous solution in a concentration of 10 mg/ml (pH 2.5 to 3) maintains > 99% of its NAD content after 8 weeks of storage at 2 to 8 °C.
The stability of an aqueous solution in a concentration of 40 mg/ml is not expected to differ from that of a solution in a concentration of 10 mg/ml.

An aqueous solution in a concentration of 50 mg/ml (pH 2.0) maintains 97% of its NAD content after 2 days, 94% after 6 days and 91% after 10 days of storage at 2 to 8 °C and about 83% after 10 days of storage at 33 °C.

An aqueous solution in a concentration of 1.38 mg/ml shows a decrease of NAD content of 4% after 8 weeks of storage at 2 to 8 °C, stored at -15 to -25 °C the stability is expected to be > 6 months. A solution of NAD in triethanolamine buffer (pH 5.0) maintians 90% of its NAD content after
10 days of storage at 2 to 8 °C and 80% after 10 days of storage at 33 °C.

Analysis Note

Absorbance coefficients: 17.6 x 103 l x mol-1 x cm-1 at 260 nm, 5.9 x 103 l x mol-1 x cm-1 at 333 nm (CN complex)
ß-Nicotinamide adenine dinucleotide hydrate cofactor (NAD) has been used in an experimental protocol, done in order to mimic the citric acid cycle on an carbon electrode, in order to increase biofuel cell energy density.[6]

Other Notes

For life science research only. Not for use in diagnostic procedures.
This is the common form of NAD.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degraded neutrophil extracellular traps promote the growth of Actinobacillus pleuropneumoniae
de Buhr N, et al.
Cell Death & Disease, 10(9), 1-16 (2019)
Daria Sokic-Lazic et al.
Biosensors & bioelectronics, 24(4), 945-950 (2008-09-09)
The citric acid cycle is one of the main metabolic pathways living cells utilize to completely oxidize biofuels to carbon dioxide and water. The overall goal of this research is to mimic the citric acid cycle at the carbon surface
Extracellular beta-nicotinamide adenine dinucleotide (beta-NAD) promotes the endothelial cell barrier integrity via PKA-and EPAC1/Rac1-dependent actin cytoskeleton rearrangement
Umapathy NS, et al.
Journal of Cellular Physiology, 223(1), 215-223 (2010)
A Mechanism of Adsorption of beta-Nicotinamide Adenine Dinucleotide on Graphene Sheets: Experiment and Theory
Pumera M, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 15(41), 10851-10856 (2009)
Experimental and theoretical electron density studies in large molecules: NAD+, beta-nicotinamide adenine dinucleotide
Guillot B, et al.
The Journal of Physical Chemistry B, 107(34), 9109-9121 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service