Skip to Content
Merck
All Photos(1)

Key Documents

XX42PRV60

Millipore

Pellicon® Cassette Low Retentate Volume Acrylic Holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151800
eCl@ss:
32031690

manufacturer/tradename

Pellicon®

Quality Level

parameter

4-50 °C temperature
50 mL min. working volume
7 bar max. inlet pressure (100 psi)
7 bar max. transmembrane pressure (100 psig)

H

25.4 cm (10 in.)

L

28 cm (11 in.)

W

19 cm (7.6 in.)

weight

11 kg (24 lb)

fitting

3/4 in. inlet/outlet sanitary flange

Related Categories

General description

Pellicon® Cassette Low Retentate Volume Acrylic Holder for Pellicon® 2 Ultrafiltration Cassettes
The Pellicon® Cassette Low Retentate Volume Acrylic Holder is perfect for anyone who needs a benchtop cassette holder versatile enough to support either microporous or ultrafiltration tangential flow filtration.

Application

Concentration, desalting or buffer exchange of:
  • Protein solutions
  • Polysaccharide solutions
  • Virus suspensions


Harvest, washing or clarification of:
  • Cell cultures and lysates
  • Colloidal suspensions
  • Viral cultures

Features and Benefits

  • Leading-edge void- free membranes to match virtually any separation challenge
  • Short flow path for higher flux and higher resolution separation capability
  • Choice of flow channel configuration providing process optimization capability
  • Predictable, fast, scale-up
  • True linear scalability from laboratory size modules to industrial assemblies for processing thousands of liters
  • Brings higher resolution, improved yields and superior back-pressure resistance
  • Ultracel membranes are composite membranes cast on a microporous substrate for defect-free membranes with superior

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service