Skip to Content
Merck
All Photos(1)

Key Documents

SX0076

Supelco

Sand

Sea

Synonym(s):

Sand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O2Si
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
11101503
EC Index Number:
238-878-4
Pricing and availability is not currently available.

form

granular

color

beige

particle size

0.2-0.8 mm

pH

5-8 (20 °C, 400 g/L in H2O)

bp

>2200 °C/1013 hPa

mp

1713 °C

shipped in

ambient

storage temp.

room temp

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Application

Sea

Analysis Note

ColorBeigeDescriptionTo pass testFormGranules, free-flowing, visibly dry

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service