Skip to Content
Merck
All Photos(1)

Documents

S25-M

Sigma-Aldrich

Mouse Serum, 100 ml

Non-sterile mouse serum collected from normal healthy mice. Used for blocking of interfering antibodies.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105901
eCl@ss:
26020100
NACRES:
NA.71

biological source

mouse

antibody form

serum

manufacturer/tradename

Chemicon®

technique(s)

blocking: suitable (interfering antibodies)

General description

Normal serum collected from healthy Kunming mice.

Total Protein:

6.77 g/dL (Biuret method)

Hemoglobin:

21.5 mg/dL

Physical form

Nonsterile liquid. Contains no preservatives. Filtered through a 0.45 micron filter system.

Storage and Stability

Store frozen at -20°C. Avoid multiple freeze-thaws.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service