Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
manufacturer/tradename
Milliflex®
input
sample type bottled water
sample type pharmaceutical(s)
sample type beer
sample type cosmetics
application(s)
bioburden testing
cosmetics
pharmaceutical
water monitoring
shipped in
ambient
General description
Support and Services
{HCompany} provides technical support and a comprehensive services offering to help you save your QC resources and facilitate the implementation of your MilliFlex Vaccum Gauge into your daily testing routine. Our highly experienced teams are there to assist you.
You can contact your local sales representative.
Application
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| application(s) bioburden testing | application(s) bioburden testing | application(s) bioburden testing | application(s) - |
| shipped in ambient | shipped in ambient | shipped in - | shipped in ambient |
| manufacturer/tradename Milliflex® | manufacturer/tradename Milliflex Oasis® | manufacturer/tradename - | manufacturer/tradename Millivac™ |
| Quality Level 400 | Quality Level SP2 | Quality Level EQ2 | Quality Level 400 |
| input sample type bottled water | input - | input - | input - |
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



