Skip to Content
Merck
All Photos(1)

Key Documents

MABT1357

Sigma-Aldrich

Anti-IZUMO1 Antibody, clone Mab120

clone Mab120, from rat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
Mab120, monoclonal
application:
IF
WB
inhibition assay
species reactivity:
mouse
technique(s):
immunofluorescence: suitable
inhibition assay: suitable
western blot: suitable
citations:
2

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Mab120, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μL

technique(s)

immunofluorescence: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Izumo1(73456)

General description

Izumo sperm-egg fusion protein 1 (UniProt: Q9D9J7; also known as Oocyte binding/fusion factor, OBF, Sperm-specific protein izumo) is encoded by the Izumo1 gene (Gene ID: 73456) in murine species. OBF is a single-pass type I membrane protein that is detected on sperm surface after the acrosome reaction. It is an essential sperm cell-surface protein required for fertilization by acting as a ligand for IZUMO1R/JUNO receptor on egg. It can be present as a monomer, homodimer, or homooligomer. It is synthesized with a signal peptide (aa 1-21), which is cleaved off in the mature form, which has an extracellular domain (aa 22-319), a short transmembrane domain (aa 320-340), and a cytoplasmic domain (aa 341-397). The extracellular domain assumes a distinct boomerang shape when not bound to IZUMO1R/JUNO. Its interaction with IZUMO1R/JUNO triggers a conformation change, so that the IZUMO1 extracellular domain assumes an upright conformation. A region comprising of 13 amino acids (148-160) is important for its interaction with IZUMO1R. Mab120 is a fusion-inhibiting antibody that inhibits fertilization. It binds to a region designated as putative functional fragment (aa 5-113). (Ref.: Inoue, N et al. (2013). Development. 140(15); 3221-3229).

Specificity

Clone Mab120 is a rat monoclonal antibody that detects Izumo sperm-egg fusion protein 1 (IZUMO1) in murine species. It targets an epitope with in the N-terminal putative functional fragment.

Immunogen

Mouse spermatozoa.

Application

Anti-IZUMO1, clone Mab120, Cat. No. MABT1357, is a rat monoclonal antibody that detects Izumo sperm-egg fusion protein 1 and has been tested for use in Immunofluorescence, Inhibition assay, and Western Blotting.
Inhibition Analysis: A representative lot inhibited fertilization in an in vitro system. (Inoue, N., et. al. (2013). Development. 140(15):3221-9).

Western Blotting Analysis: A representative lot detected IZUMO1 in Western Blotting applications (Inoue, N., et. al. (2013). Development. 140(15):3221-9).

Immunofluorescence Analysis: A representative lot detected IZUMO1 in Immunofluorescence applications (Inoue, N., et. al. (2013). Development. 140(15):3221-9).
Research Category
Cell Structure

Quality

Evaluated by Western Blotting in mouse testis tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected IZUMO1 in mouse testis tissue lysate.

Target description

~56 kDa observed; 44.88 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas G Brukman et al.
eLife, 13 (2024-01-24)
The fusion of mammalian gametes requires the interaction between IZUMO1 on the sperm and JUNO on the oocyte. We have recently shown that ectopic expression of mouse IZUMO1 induces cell-cell fusion and that sperm can fuse to fibroblasts expressing JUNO.
Nicolas G Brukman et al.
The Journal of cell biology, 222(2) (2022-11-18)
Mammalian sperm-egg adhesion depends on the trans-interaction between the sperm-specific type I glycoprotein IZUMO1 and its oocyte-specific GPI-anchored receptor JUNO. However, the mechanisms and proteins (fusogens) that mediate the following step of gamete fusion remain unknown. Using live imaging and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service