Skip to Content
Merck
All Photos(1)

Key Documents

MABS809

Sigma-Aldrich

Anti-JAK2, clone 1C1 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
1C1
monoclonal
application:
WB
species reactivity:
mouse, human
technique(s):
western blot: suitable
citations:

antibody form

ascites fluid

Quality Level

clone

1C1
monoclonal

purified by

(unpurified)

species reactivity

mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

General description

Purified recombinant fragment of JAK2 expressed in E. Coli

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis (WB): A 1:500-2,000 dilution of this antibody detected JAK2 in THP-1 cell lysate.

Physical properties

~129 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse monoclonal IgG1ascitic fluid containing 0.03% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service