Skip to Content
Merck
All Photos(1)

Key Documents

MABN2425

Sigma-Aldrich

Anti-TRPV2 Antibody, clone 17A11

clone 17A11, from mouse

Synonym(s):

Transient receptor potential cation channel subfamily V member 2, Osm-9-like TRP channel 2, OTRPC2, Stretch-activated channel 2B, Vanilloid receptor-like protein 1, VRL-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

17A11, monoclonal

species reactivity

mouse, rat

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Trpv2(22368)
rat ... Trpv2(29465)

General description

Transient receptor potential cation channel subfamily V member 2 (UniProt: Q9WUD2; also known as TrpV2, Osm-9-like TRP channel 2, OTRPC2, Stretch-activated channel 2B, Vanilloid receptor-like protein 1, VRL-1) is encoded by the Trpv2 (also known as Sac2b, Vrl1) gene (Gene ID: 29465) in rat. TrpV2 is an ubiquitously expressed, calcium-permeable, non-selective cation channel with an outward rectification. It has been implicated in various physiological and pathological processes, including growth factor signaling, macrophage migration, cancer cell metastasis, heart disease, and neuronal cell development. It appears to be regulated by growth factors, like IGF1, PDGF, and morphogenetic neuropeptide/head activator. It is also activated by temperatures higher than 52 degree C, but is not activated by vanilloids and acidic pH. TrpV2 channel is upregulated by the nerve growth factor (NGF) via the mitogen-activated protein kinase (MAPK) signaling pathway to augment neurite outgrowth. Although NGF increases TrpV2 content, it does not induce the translocation of TrpV2 from cytoplasm to cell surface in PC12 cells. (Ref.: Cohen MR., et al. (2015). Mol. Cell Biol. 35(24): 4238-4252).

Specificity

Clone 17A11 detects Transient receptor potential cation channel subfamily V member 2 (TRPV2) in mouse and rat cells.

Immunogen

1D4-tagged full length rat Transient receptor potential cation channel subfamily V member 2 (TRPV2).

Application

Anti-TRPV2, clone 17A11, Cat. No. MABN2425, is a highly specific mouse monoclonal antibody that targets TRPV2 and has been tested for use in Immunocytochemistry and Western Blotting.
Western Blotting Analysis: A representative lot detected TRPV2 in Western Blotting applications (Cohen, M.R., et. al. (2013). PLoS One. 8(12):e85392).

Immunocytochemistry Analysis: A representative lot detected TRPV2 in Immunocytochemistry applications (Cohen, M.R., et. al. (2013). PLoS One. 8(12):e85392; Cohen M.R., et. al. (2015). Mol Cell Biol. 35(24):4238-52).

Quality

Evaluated by Immunocytochemistry in PC12 cells treated with NGF.

Immunocytochemistry Analysis: 2-10 µg/mL of this antibody detected TRPV2 in PC12 cells treated with mouse NGF.

Target description

86.71 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service