Skip to Content
Merck
All Photos(1)

Key Documents

MABN2263

Sigma-Aldrich

Anti-ZMYM2 Antibody, clone 9F12.1

clone 9F12.1, from mouse

Synonym(s):

Zinc finger MYM-type protein 2, Fused in myeloproliferative disorders protein, Rearranged in atypical myeloproliferative disorder protein, Zinc finger protein 198

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9F12.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ZMYM2(7750)

Related Categories

General description

Zinc finger MYM-type protein 2 (UniProt: Q9UBW7; also known as Fused in myeloproliferative disorders protein; Rearranged in atypical myeloproliferative disorder protein; Zinc finger protein 198) is encoded by the ZMYM2 (also known as FIM, RAMP, ZNF198) gene (Gene ID: 7750) in human. Zinc finger MYM-type protein 2 is a nuclear protein that may function as a transcription factor. Two isoforms of Zinc finger MYM-type protein 2 have been described that are produced by alternative splicing. The SUMO-interacting motifs (SIM) of Zinc finger MYM-type protein 2 are shown to be essential for its recruitment to chromatin. The SIM motifs play an important role in its gene-regulatory activities, however, they are not required for the intrinsic transcriptional repressive properties of Zinc finger MYM-type protein 2. A chromosomal aberration involving ZMYM2 is reported to cause stem cell leukemia lymphoma syndrome (SCLL), which is characterized as lymphoblastic lymphoma in association with a myeloproliferative disorder accompanied by pronounced peripheral eosinophilia and/or prominent eosinophilic infiltrates in the affected bone marrow. (Ref.: Aguilar-Martinez, E., et al. (2015). Proc. Natl. Acad. Sci. USA. 112 (35); E4854-E4863).

Specificity

Clone 9F12.1 detects Zinc finger MYM-type protein 2 in human cells. It targets an epitope within 167 amino acids from the C-teminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 167 amino acids from the C-terminal region of human Zinc finger MYM-type protein 2.

Application

Anti-ZMYM2, clone 9F12.1, Cat. No. MABN2263, is a highly specific mouse monoclonal antibody that targets Zinc finger MYM-type protein 2 and has been tested in Western Blotting.

Quality

Evaluated by Western Blotting in human hippocampus tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected ZMYM2 in human hippocampus tissue lysate.

Target description

~155 kDa observed; 154.91 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service