Skip to Content
Merck
All Photos(4)

Documents

MABN1101

Sigma-Aldrich

Anti-SRY, clone 1G4 Antibody

clone 1G4, from mouse

Synonym(s):

Sex-determining region Y protein, Testis-determining factor, SRY

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G4, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SRY(6736)

General description

Sex-determining region Y protein, also known as Testis-determining factor, and encoded by the human gene SRY or TDF is a transcriptional regulator that controls a genetic switch in male development. SRY is necessary and sufficient for initiating male sex determination by directing the development of supporting cell precursors (pre-Sertoli cells) to Sertoli rather than granulosa cells. In male adult brain SRY is involved in the maintenance of motor functions of dopaminergic neurons. SRY is also involved in different aspects of gene regulation including promoter activation or repression, DNA bending and manipulation. The SRY HMG box recognizes DNA by partial intercalation into the minor groove, and SRY is also involved in pre-mRNA splicing. SRY is expressed in the developing testis and surrounding tissue and can be found expressed in neurons in the adult. SRY is nuclear associated.

Immunogen

Purified recombinant fragment of human SRY expressed in E. Coli

Application

Anti-SRY, clone 1G4 Antibody is an antibody against SRY for use in western blotting, IHC, flow cytometry & ELISA.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected SRY in cervical and ovarian cancer tissues.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected SRY in HepG2 cells.

ELISA: A representative lot detected control Antigen (100 ng) and Antigen (10 ng, 50 ng, 100 ng) in ELISA (Direct).
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in NTERA-2 cell lysate.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected SRY in NTERA-2 cell lysate.

Target description

~36 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal in PBS containing up to 0.1% sodium azide and 0.5% protein stabilizer.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugene Lee et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 37(7), 1382-1399 (2022-04-25)
Poor survival of grafted cells is the major impediment of successful cell-based therapies for bone regeneration. Implanted cells undergo rapid death in an ischemic environment largely because of hypoxia and metabolic stress from glucose deficiency. Understanding the intracellular metabolic processes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service