Skip to Content
Merck
All Photos(2)

Key Documents

MABF950

Sigma-Aldrich

Anti-Integrin alpha-4 (CD49d) Antibody, clone TA-2

clone TA-2, from mouse

Synonym(s):

Protein Itga4, CD49d, IA4, VLA-4 alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TA-2, monoclonal

species reactivity

rat

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Integrin alpha-4 (UniProt: D3ZMQ3; also known as CD49d, Protein Itga4) is encoded by the Itga4 gene in rat. Integrin-Alpha 4 contains 7 GF-GAP repeats. Unlike other integrin alpha chains, alpha 4 neither contains an I-domain, nor undergoes disulfide-linked cleavage. It associates with either beta-1 or beta-7 to generate a heterodimer. The alpha subunit can sometimes be cleaved into two non-covalently associated fragments. Integrin alpha-4/beta-1 (alpha 4/CD29; VLA-4) may regulate leukocyte to endothelial cells adhesion by controlling Junctional adhesion molecules-L homodimerization. Integrins alpha-4/beta-1 and alpha-4/beta-7 are receptors for fibronectin and they recognize one or more domains within the alternatively spliced CS-1 and CS-5 regions of fibronectin. Integrin alpha-4/beta-1 plays a major role in the regulation of immune cell recruitment to inflamed endothelia and sites of inflammation. On activated endothelial cells integrin alpha-4/beta-1 triggers homotypic aggregation for most alpha-4/beta-1-positive leukocyte cell lines. This monoclonal antibody (clone TA-2) is shown to inhibit rat lymphocyte adhesion to stimulated HUVEC. It also blocks adhesion small peritoneal exudate lymphocytes (sPEL) adhesion to TNF-alpha and IL-1 stimulated human umbilical vein endothelial cells. (Ref.: Issekutz, TB., and Wykretowicz, A (1991). J. Immunol. 147:109-116).

Specificity

Clone TA-2 detects CD49d/Integrin alpha-4 in Rat splenocytes and lymphocytes.

Immunogen

Small peritoneal exudate lymphocytes (sPELs).

Application

Flow Cytometry Analysis: A representative lot detected Integrin alpha-4 (CD49d) in Flow Cytometry appications (Latta, M., et. al. (2007). Immunology. 121(4):555-64).

Flow Cytometry Analysis: A representative lot detected Integrin alpha-4 (CD49d) in Flow Cytometry appications (Schlager, C., et. al. (2016). Nature. 530(7590):349-53).

Agonist or Inhibitor Analysis: A representative lot detected Integrin alpha-4 (CD49d) in Agonist or Inhibitor appications (Issekutz, T.B., et. al. (1991). J Immunol. 147(12):4178-84).

Agonist or Inhibitor Analysis: A representative lot detected Integrin alpha-4 (CD49d) in Agonist or Inhibitor appications (Odoardi, F., et. al. (2012). Nature. 488(7413):675-9).

Agonist or Inhibitor Analysis: A representative lot detected Integrin alpha-4 (CD49d) in Agonist or Inhibitor appications (Schlager, C., et. al. (2016). Nature. 530(7590):349-53).

Immunoprecipitation Analysis: A representative lot detected Integrin alpha-4 (CD49d) in Immunoprecipitation appications (Schlager, C., et. al. (2016). Nature. 530(7590):349-53).
This mouse monoclonal Anti-Integrin alpha-4 (CD49d), clone TA-2, Cat. No. MABF950 is tested for use in Flow Cytometry, Immunoprecipitation, and in functional studies as Agonist or Inhibitor for the detection of CD49d/Integrin alpha-4.

Quality

Evaluated by Flow Cytometry in rat splenocytes and lymphocytes.

Flow Cytometry Analysis: 1 µg of this antibody detected Integrin alpha-4 (CD49d) in one million rat splenocytes and lymphocytes.

Target description

115.59 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service