Skip to Content
Merck
All Photos(2)

Key Documents

MABC632

Sigma-Aldrich

Anti-MAP3K5 Antibody, clone 2E4

ascites fluid, clone 2E4, from mouse

Synonym(s):

Mitogen-activated protein kinase kinase kinase 5, Apoptosis signal-regulating kinase 1, ASK-1, MAPK/ERK kinase kinase 5, MEK kinase 5, MEKK 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2E4, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAP3K5(4217)

Related Categories

General description

Mitogen-activated protein kinase kinase kinase 5 (MAP3K5), also known as, MAPK/ERK kinase 5 (MEK Kinase 5; MEKK 5) or Apoptosis signal-regulating kinase 1 (ASK-1) protein, is a serine/threonine kinase that is part of the MAP kinase signal transduction pathway. The MAP kinase pathway is central to several signaling cascades the help determine everything from cell fate in differentiation to cell survival in apoptosis. MAP3K5 is critical for innate immune response against numerous pathogens and MAP3K5 also mediates cells responses to various cellular stresses such as oxidative stress, and responses mediated by inflammatory signaling agents such as TNF or LPS. Moreover, MAP3K5 has has been found to be involved in cancer, diabetes, cardiovascular and neurodegenerative diseases. MAP3K5 is membrane associated though not membrane bound directly. Its expression thus is primarily endoplasmic reticulum but can become strongly perinuclear under some conditions. Found in most tissues, it is abundant in heart and pancreas. It′s activity is regulated via phosphorylation-dephosphorylation processes as well as though its interactions with a many other protein partners to form numerous multimolecular cell signaling complexes. EMD-Millipore’s Anti-MAP3K5 mouse monoclonal, clone 2E4, has been tested in western blotting against recombinant human MAP3K5 protein, and in paraffin-embedded immunohistochemistry with human cancer tissues, and in immunofluorescent immunocytochemistry, and flow cytometry on human HeLa cells in culture.

Immunogen

Purified recombinant fragment of human MAP3K5 expressed in E. coli.

Application

Anti-MAP3K5 Antibody, clone 2E4 is a highly specific mouse monoclonal antibody, that targets MAP3K5 & has been tested in western blotting, IHC, ICC, Flow Cytometry & ELISA.
Immunohistochemistry Analysis: A 1:100-1,000 dilution from a representative lot detected MAP3K5 in human pancreas, heart, breast cancer, and prostate tissues.

Immunocytochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected MAP3K5 in HeLa cells.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected MAP3K5 in HeLa cells.

ELISA: A representative lot detected MAP3K5 in ELISA (Direct).

Optimal working dilutions must be determined by end user
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MAP3K5 in 10 µg of HepG2 cell lysate.

Target description

~155 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HepG2 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norman L Lehman et al.
Nature communications, 13(1), 2083-2083 (2022-04-21)
Astroblastomas (ABs) are rare brain tumors of unknown origin. We performed an integrative genetic and epigenetic analysis of AB-like tumors. Here, we show that tumors traceable to neural stem/progenitor cells (radial glia) that emerge during early to later brain development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service