Skip to Content
Merck
All Photos(1)

Key Documents

MABC1832

Sigma-Aldrich

Anti-CD44v4 Antibody, clone C44Mab-108

Synonym(s):

CDw44, Epican, Extracellular matrix receptor III (ECMR-III) GP90 lymphocyte homing/adhesion receptor, HUTCH-I, Heparan sulfate proteoglycan, Hermes antigen, Hyaluronate receptor, LHR, Phagocytic glycoprotein 1 (PGP-1), Phagocytic glycoprotein I (PGP-I)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
clone:
C44Mab-108, monoclonal
application:
ELISA
FACS
IHC
WB
species reactivity:
human
technique(s):
ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

C44Mab-108, monoclonal

mol wt

calculated mol wt 82 kDa
observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... CD44(960)

Specificity

Clone C44Mab-108 is a mouse monoclonal antibody that detects CD44v4. It targets an epitope within 19 amino acids from the N-terminal extracellular domain of the protein.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal extracellular domain of human CD44v4.

Application

Quality Control Testing

Evaluated by Flow Cytometry in a CHO cells overexpressing CD44v3-10.

Flow Cytometry Analysis: 1.0 µg of this antibody detected CD44v6 in a one million CHO cells overexpressing CD44v3-10.

Tested Applications

ELISA Analysis: A representative lot detected CD44v4 in ELISA applications (Suzuki, H., et al. (2023). Curr. Issues Mol. Biol. 45(3):1875-1888).

Western Blotting Analysis: A representative lot detected CD44v4 in Western Blotting applications (Suzuki, H., et al. (2023). Curr. Issues Mol. Biol. 45(3):1875-1888).

Flow Cytometry Analysis: A representative lot detected CD44v4 in Flow Cytometry applications (Suzuki, H., et al. (2023). Curr Issues Mol Biol. 45(3):1875-1888).

Immunohistochemistry Applications: A representative lot detected CD44v4 in Immunohistochemistry applications (Suzuki, H., et al. (2023). Curr. Issues Mol. Biol. 45(3):1875-1888).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

CD44 antigen (UniProt: P16070; also known as CDw44, Epican, Extracellular matrix receptor III (ECMR-III), GP90 lymphocyte homing/adhesion receptor, HUTCH-I, Heparan sulfate proteoglycan, Hermes antigen, Hyaluronate receptor, Phagocytic glycoprotein 1 (PGP-1), Phagocytic glycoprotein I (PGP-I)) is encoded by the CD44 (also known as LHR, MDU2, MDU3, MIC4) gene (Gene ID: 960) in human. CD44 is a Type 1 transmembrane glycoprotein involved in cell adhesion, migration, and proliferation. It also plays a role in lymphocyte activation, tumor metastasis, and wound healing. It is encoded by the CD44 gene, and it is expressed in a wide range of cell types. The protein′s structure includes a hyaluronic acid (HA) binding domain, a stem region, and a variable extracellular domain. Alternative splicing may produce 19 isoforms. CD44 splice variants (CD44v) play a role in tumorigenesis and malignancy. It is also implicated in various other diseases, including autoimmune disorders, cardiovascular diseases and fibrosis.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service