Skip to Content
Merck
All Photos(2)

Key Documents

MAB2160

Sigma-Aldrich

Anti-Fragile X Mental Retardation Protein Antibody, clone 1C3

ascites fluid, clone 1C3, Chemicon®

Synonym(s):

FMRP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1C3, monoclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

suitability

not suitable for flow cytometry
not suitable for immunoprecipitation

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

Fragile X Mental Retardation Protein (FMRP) is a RNA-binding protein that is associated to polysomes and may be involved in the transport of mRNA from the nucleus to the cytoplasm. Defects in FMR1 are the cause of Fragile X syndrome, which is a common genetic disease characterized by moderate to severe mental retardation, macroorchidism, large ears, prominent jaw, and high-pitched, jocular speech. The defect in most fragile X syndrome patients results from an amplification of a CGG repeat region which is directly in front of the coding region.

Specificity

Human and mouse FMRP. The epitope is localized in the N-terminal half of FMRP. Cross reaction with FXR protein may be detected in cases of high expression of the latter proteins.

Immunogen

Fusion protein with a full length FMRP (human).

Application

Anti-Fragile X Mental Retardation Protein Antibody, clone 1C3 detects level of Fragile X Mental Retardation Protein & has been published & validated for use in ELISA, IC, IH, IH(P) & WB.
Immunohistochemistry:
Frozen and paraffin sections. 1:500-1:5,000 dilution of a previous lot was used.

ELISA:
A 1:500-1:5,000 dilution of a previous lot was used in ELISA.

Detection of FMRP on Blood Smears:
A 1:500-1:5,000 dilution of a previous lot was used.

Immunocytochemistry:
A 1:500-1:5,000 dilution of a previous lot was used on transfected cells. Light fixation (2% PFA, permeabilize with 0.1% triton in block only)

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

RNA Binding Protein (RBP)

Quality

Evaluated by Western Blot on Mouse E17 spinal cord lysates.

Western Blotting Analysis:
1:500 dilution of this antibody detected Fragile X Mental Retardation Protein on 10 μg of Mouse E17 spinal cord lysates.

Target description

~71 kDa

Physical form

Ascites mouse monoclonal IgG1k fluid containing no preservatives
Unpurified

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
HeLa whole cell lysate, HeLa nuclear lysate, mouse E17 spinal cord lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The RNA-binding protein fragile X-related 1 regulates somite formation in Xenopus laevis.
Huot, ME; Bisson, N; Davidovic, L; Mazroui, R; Labelle, Y; Moss, T; Khandjian, EW
Molecular Biology of the Cell null
Gabi Schutzius et al.
Journal of neurodevelopmental disorders, 5(1), 8-8 (2013-04-04)
Hypermethylation of the fragile X mental retardation 1 gene FMR1 results in decreased expression of FMR1 protein FMRP, which is the underlying cause of Fragile X syndrome - an incurable neurological disorder characterized by mental retardation, anxiety, epileptic episodes and
FMRP S499 is phosphorylated independent of mTORC1-S6K1 activity.
Bartley, CM; O'Keefe, RA; Bordey, A
Testing null
Alice Filippini et al.
RNA biology, 14(11), 1580-1591 (2017-06-24)
The fragile X syndrome (FXS), the most common form of inherited intellectual disability, is due to the absence of FMRP, a protein regulating RNA metabolism. Recently, an unexpected function of FMRP in modulating the activity of Adenosine Deaminase Acting on
Julie B Zang et al.
PLoS genetics, 5(12), e1000758-e1000758 (2009-12-17)
The mental retardation, autistic features, and behavioral abnormalities characteristic of the Fragile X mental retardation syndrome result from the loss of function of the RNA-binding protein FMRP. The disease is usually caused by a triplet repeat expansion in the 5'UTR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service