Skip to Content
Merck
All Photos(1)

Key Documents

MAB1501X

Sigma-Aldrich

Anti-Actin Antibody, clone C4, Alexa Fluor 488 conjugated

clone C4, Chemicon®, from mouse

Synonym(s):

MAB1501, MAB1501R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
conjugate:
ALEXA FLUOR 488
application:
IF
clone:
C4, monoclonal
species reactivity:
human
citations:
2
technique(s):
immunofluorescence: suitable

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

C4, monoclonal

species reactivity

human

species reactivity (predicted by homology)

chicken (based on 100% sequence homology)

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2bκ

NCBI accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACTA1(58)

General description

Actins are ubiquitous eukaryotic proteins that serve as a multi-functional, basic building blocks of cytoskeletal microfilaments. They play critical roles in a wide range of cellular processes, including cell division, cell migration, chromatin remodeling, trascriptional regulation and vesicle trafficking. These funstions are attributed to their ability to form filaments, which can quickly assemble and disassemble depending upon the needs of the cell. At least six different actin types have been reported in mammals. Although actins show about 90% overall sequence homology, isoforms do not show spatial, temporal and tissue-specific expression patterns and only 50-60% homology is found in their 18 N-terminal residues. Beta and gamma-actins, also known as cytoplasmic actins, are highly conserved in higher animals and are predominantly expressed in non-muscle cells where they control cell structure. Exocytosis, and motility. They are nearly identical proteins and differ only in four amino acids at the N-terminal region. The other four actin isoforms are typically found in specific adult muscle tissue types. Alpha-cardiac and alpha-skeletal actins are expressed in striated cardiac and skeletal muscles, respectively. Alpha and gamma actins are primarily found in vascular smooth muscle and enteric smooth muscles, respectively. It has been shown that under calcium-bound conditions, beta-actin exhibits more dynamic behavior than gamma-actin with higher rates of polymerization and depolymerization. Also, beta- and gamma-actins can readily copolymerize, and the resulting filaments exhibit polymerization and depolymerization rates that vary depending on the ration of beta- to gamma-actin (Lessard, JL.,et al.(1988). Cell Motility Cytoskeleton 10(3); 349-362.

Specificity

MAB1501X is a pan-actin antibody that binds to an epitope in a highly conserved region of actin; therefore, this antibody reacts with all six isoforms of vertabrate actin (Lessard, 1988). Reacts with both globular (G) and fillimentous (F) forms of actin and does not interfere with actin polymerization to form filaments, at a ratio as high as one antibody per two actin monomers. However, this antibody does increase the extent of polymerization when used at a lower ratio of antibody to actin. In addition to labeling myotubes, anti-actin stains myoblasts and fibroblasts. Although clone C4 is prepared as an antibody to chicken gizzard muscles actin, it reacts with actins from all vertebrates, as well as with Dictyostelium discoideum and Physarum polycephalum actins (Lessard, 1988).
Other species have not been tested.

Immunogen

Epitope is conserved in all known actins.
Purified chicken gizzard actin (Lessard, 1988).

Application

Anti-Actin Antibody, clone C4, Alexa Fluor 488 conjugated detects level of Actin & has been published & validated for use in IF.
Immunofluorescence: 1:200-1:1000

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluatied on a representative lot by immunofluorescence using HeLa cells.

Target description

~43 kDa

Linkage

Replaces: MAB1501X

Physical form

Protein G Purified
Purified in PBS with 15 mg/mL BSA and 0.05% NaN3, conjugated to Alexa Fluor 488.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 1 year after date of receipt.

Analysis Note

Control
HeLa cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Monici et al.
Molecular bioSystems, 9(6), 1147-1161 (2013-02-01)
Laser therapy is used in physical medicine and rehabilitation to accelerate muscle recovery and in sports medicine to prevent damages produced by metabolic disturbances and inflammatory reactions after heavy exercise. The aim of this research was to get insight into
Shirley Genah et al.
Biomedicines, 9(3) (2021-04-04)
The fine control of inflammation following injury avoids fibrotic scars or impaired wounds. Due to side effects by anti-inflammatory drugs, the research is continuously active to define alternative therapies. Among them, physical countermeasures such as photobiomodulation therapy (PBMT) are considered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service