HX0291
Hexane, mixture of isomers
≥95% (n-hexane basis), ≥98.5% (hexane isomers + methylcyclopentane), suitable for HPLC, meets USP/NF
Synonym(s):
Hexane, n-Hexane
About This Item
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)
Recommended Products
Product Name
n-Hexane 95%, HPLC, Meets Reagent Specifications for testing USP/NF monographs
grade
reagent
Quality Level
Agency
meets USP/NF
vapor density
~3 (vs air)
vapor pressure
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)
Assay
≥95% (n-hexane basis)
≥98.5% (hexane isomers + methylcyclopentane)
form
liquid
autoignition temp.
453 °F
expl. lim.
7.7 %
technique(s)
HPLC: suitable
impurities
≤0.0003 meq/g Water-soluble titrable acid
≤0.005% Sulfur compounds (as S)
≤0.01% Water (H2O)
evapn. residue
≤5 ppm
color
APHA: ≤10
refractive index
n20/D 1.375 (lit.)
bp
69 °C (lit.)
mp
−95 °C (lit.)
transition temp
flash point -22 °C
solubility
0.0095 g/L
density
0.659 g/mL at 25 °C (lit.)
UV cutoff
≤195 nm
UV absorption
λ: 195 nm Amax: ≤1.00
λ: 210 nm Amax: ≤0.30
λ: 220 nm Amax: ≤0.10
λ: 230 nm Amax: ≤0.05
λ: 240 nm Amax: ≤0.04
λ: 250 nm Amax: ≤0.02
λ: 280-400 nm Amax: ≤0.01
shipped in
ambient
storage temp.
room temp
SMILES string
CCCCCC
InChI
1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3
InChI key
VLKZOEOYAKHREP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Analysis Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system, Nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-7.6 °F - closed cup
Flash Point(C)
-22 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service