Skip to Content
Merck
All Photos(1)

Key Documents

GX0190

Supelco

Glycerol

Synonym(s):

Glycerin, 1, 1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-289-5
Assay:
≥95%
technique(s):
electrophoresis: suitable
bp:
182 °C/20 mmHg (lit.)
vapor pressure:
<1 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3.1 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥95%

form

liquid

autoignition temp.

698 °F

technique(s)

electrophoresis: suitable

impurities

≤0.001% Chloride (Cl)
≤0.002% Sulfate (SO4)
≤0.003% Chlorinated compounds (as Cl)
≤5 ppm Heavy metals (as Pb)

refractive index

n20/D 1.474 (lit.)

pH

5 (20 °C, 100 g/L in H2O)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

transition temp

flash point 199 °C

density

1.25 g/mL (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Acrolein, glucose, and ammonium compoundsTo pass testAssay95% minChloride (Cl)0.001% maxChlorinated compounds (as Cl)0.003% maxColorTo pass testDescriptionTo pass testFatty acids and estersTo pass testHeavy metals (as Pb)5ppm maxIdentificationTo pass testReadily carbonizable substancesTo pass testResidue after ignition0.01% maxSpecific Gravity at 25/25°C1.249 minSulfate (SO4)0.002% max

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service