GREL5
MPAGE®® Gel Releasers
for use with mPAGE® Gel Caster and mPAGE® Mini Gel Tank
About This Item
Recommended Products
Product Name
mPAGE® Gel Releasers, 5pk, for use with mPAGE® Gel Caster and mPAGE® Mini Gel Tank
packaging
pkg of 5 packs
manufacturer/tradename
Millipore
technique(s)
SDS-PAGE: suitable
electrophoresis: suitable
compatibility
for use with mPAGE® Gel Caster
General description
The mPAGE® Gel Caster offers a leak-free solution for handcasting up to 2 mini protein gels. Gels cast
are 8 cm x 10 cm with a gel thickness of 1.0 mm, 0.75 mm, or 1.5 mm, along
with 10 or 15-well options for sample loading. The mPAGE® Gel Caster has been
designed to simplify hand casting gels and to eliminate leakage during the casting
process. The gel caster is available as a standalone device (GCR2) or as a gel casting device kit for casting mini gels gels with 0.75 mm, 1.0 mm and 1.5 mm thickness (MGCK-75M, MGCK-10M, MGCK-15M).
Legal Information
related product
Protocols
Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.
Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.
Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.
Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.