Skip to Content
Merck
All Photos(2)

Key Documents

EZGSWG474

Millipore

EZ-Pak® Membrane Filter

mixed cellulose esters (MCE), pore size 0.22 μm, filter diam. 47 mm, gridded white filter, suitable for bioburden testing

Synonym(s):

Membrane filters, MCE filters, GSWG EZ-PAK, Semi-permeable membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

gridded filter
gridded white filter
mixed cellulose esters (MCE)

Quality Level

Agency

ISO 7704

sterility

sterile; ethylene oxide treated

feature

easily wetted with water
hydrophilic
individually wrapped

manufacturer/tradename

EZ-Pak®

filter diam.

47 mm

matrix

MF-Millipore

pore size

0.22 μm pore size

input

sample(s) (environmental)
sample type beer
sample type pharmaceutical(s)
sample type cider
sample type soft drink
sample type bottled water
sample type wine
sample type sport drink
sample type beverage(s)
sample type food(s)
sample type cosmetics

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical

compatibility

for use with EZ-Fit® Filtration Unit

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

EZ-Pak® Filters are made of Mixed cellulose esters (MCE) with 022 µm pore size, white, gridded and 47mm diameter. These filters possess high recoveries and are more appropriate for general microbiological testing. EZ-Pak® membranes and dispenser revolutionize filter handling for Membrane Filtration (MF) method testing by combining the benefits of ready-to-use, sterile, individually packed membranes with unique handling ease using the EZ Pak® Curve dispenser for dispensing.

Application

The EZ-Pak® system is suitable for testing drinking, bottled and mineral water according to international standards. These filters find application in Clinical, Cosmetics, Environmental, Food and Beverage, Industrial QC and Pharmaceutical industry. Microbial analysis of beer, bottled water, cider, sport drink and wine by membrane filtration method

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatemeh Gholami et al.
Environmental science and pollution research international, 26(34), 35218-35226 (2019-11-07)
Oxygen-releasing compounds (ORCs) have recently gained much attention in contaminated groundwater remediation. We investigated the impact of calcium peroxide nanoparticles on the groundwater indigenous bacteria in a bioremediation process by permeable reactive barrier (PRB). Three sand-packed columns were applied, including

Related Content

A touch-free membrane filter dispenser can streamline your membrane filtration testing workflow. Use with EZ-Pak® filter membranes for high-volume sampling.

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service