Skip to Content
Merck
All Photos(1)

Documents

EZFITMVG3

Millipore

EZ-Fit® filtration head gasket

EZ-Fit® filtration head, suitable for (General microbial analysis)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

silicone

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 3 units

manufacturer/tradename

EZ-Fit®

input

sample type pharmaceutical(s)
sample type beer
sample type bottled water
sample type cosmetics

suitability

suitable for (General microbial analysis)

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

General description

EZ-Fit® filtration head gasket is a spare for the EZ-Fit® filtration head and ensure optimal gripping of the filtration head while filtering.

Application

EZ-Fit® filtration head gasket is a EZ-Fit® Manifold accessory for the filtration of the test samples for general microbiological analysis, bioburden testing, & water monitoring.

Preparation Note

Sterilization Method
Autoclavable

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service